Search
Search

LCMT2| Gene Symbol: LCMT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014793.4 | NP_055608.2 | 1 | 2121 | 98 | |
| GenBank mRNA | AF265443.1 | - | 1 | 1614 | 98 | |
| AK292587.1 | - | 1 | 2047 | 98 | ||
| AK300820.1 | - | 2 | 1330 | 98 | ||
| BC015949.2 | - | 1 | 2104 | 3922888 | 98 | |
Set Membership: |
|