Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs07289533_m1
          See other TGIF1 GE Assays ›
          Gene Symbol:
          TGIF1
          Gene Name:
          TGFB induced factor homeobox 1
          Gene Aliases:
          HPE4, TGIF
          Chromosome Location:
          Chr.18: 3411927 - 3458412 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs07289533_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: TGIF1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001278682.1 NP_001265611.1 2 - 3 463 92
          NM_001278684.1 NP_001265613.1 3 - 4 820 92
          NM_001278686.1 NP_001265615.1 3 - 4 486 92
          NM_003244.3 NP_003235.1 2 - 3 553 92
          NM_170695.3 NP_733796.2 2 - 3 1197 92
          NM_173207.2 NP_775299.1 2 - 3 431 92
          NM_173208.2 NP_775300.1 3 - 4 823 92
          NM_173209.2 NP_775301.1 2 - 3 639 92
          NM_173210.2 NP_775302.1 2 - 3 590 92
          NM_173211.1 NP_775303.1 2 - 3 302 92
          NM_174886.2 NP_777480.1 3 - 4 620 92
          XM_011525735.2 XP_011524037.1 2 - 3 799 92
          XM_017025958.1 XP_016881447.1 3 - 4 884 92
          XM_017025959.1 XP_016881448.1 2 - 3 669 92
          GenBank mRNA AF179900.1 - 2 - 3 241 92
          AI813434.1 - 3 - 4 441 92
          AK026766.1 - 1 - 2 1159 92
          AK291112.1 - 2 - 3 387 92
          BC000814.1 - 2 - 3 319 3453337 92
          BC031268.1 - 2 - 3 925 5284849 92
          BE296707.1 - 2 - 3 315 92
          BE906361.1 - 3 - 4 472 92
          BF244060.1 - 2 - 3 91
          BF446044.1 - 3 - 4 435 92
          BM849453.1 - 3 - 4 489 92
          BQ437864.1 - 2 - 3 551 92
          BU657748.1 - 2 - 3 407 92
          BX952985.1 - 2 - 3 320 92
          CB161219.1 - 2 - 3 463 92
          CB996365.1 - 3 - 4 647 92
          CR450333.1 - 2 - 3 235 92
          D82120.1 - 2 - 3 302 92
          DR007147.1 - 2 - 3 643 92
          X89750.1 - 2 - 3 546 92

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene TGIF1

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          multicellular organism development
          response to drug
          cellular response to growth factor stimulus
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          co-SMAD binding

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline