Search
Search

C2orf27B| Gene Symbol: C2orf27B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214461.2 | NP_999626.1 | 2 - 3 | 534 | 63 | |
| GenBank mRNA | BC043584.1 | - | 2 - 3 | 534 | 5229449 | 63 |
Set Membership: |
|