Search
Search

TEX22| Gene Symbol: TEX22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195082.1 | NP_001182011.1 | 2 - 3 | 244 | 72 | |
| XM_006720234.3 | XP_006720297.1 | 2 - 3 | 518 | 72 | ||
| GenBank mRNA | BX378619.2 | - | 2 - 3 | 244 | 72 | |
Set Membership: |
|