Search
Search

DHRS4L2| Gene Symbol: DHRS4L2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193637.1 | NP_001180566.1 | 3 - 4 | 737 | 108 | |
| GenBank mRNA | DN237881.1 | - | 3 - 4 | 424 | 108 | |
Set Membership: |
|