Search
Search
| Gene Symbol: GYPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002102.3 | NP_002093.2 | 2 - 3 | 193 | 90 | |
| NM_198682.2 | NP_941391.2 | 2 - 3 | 193 | 90 | ||
| XM_017008139.1 | XP_016863628.1 | 3 - 4 | 4120 | 90 | ||
| XM_017008140.1 | XP_016863629.1 | 3 - 4 | 4120 | 90 | ||
| GenBank mRNA | BC017864.1 | - | 2 - 3 | 189 | 4686481 | 90 |
| BC064396.1 | - | 2 - 3 | 150 | 90 | ||
| BG528203.1 | - | 2 - 3 | 189 | 90 | ||
| BX091225.1 | - | 2 - 3 | 161 | 90 | ||
| M29610.1 | - | 2 - 3 | 193 | 90 | ||
Set Membership: |
|