Search
Search
| Gene Symbol: JPH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271605.1 | NP_001258534.1 | 2 | 818 | 77 | |
| XM_017023483.1 | XP_016878972.1 | 2 | 749 | 77 | ||
| GenBank mRNA | BC008690.1 | - | 2 | 234 | 77 | |
Set Membership: |
|