Search
Search

SMR3A| Gene Symbol: SMR3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012390.3 | NP_036522.3 | 3 | 219 | 82 | |
| XM_017008021.1 | XP_016863510.1 | 2 | 190 | 82 | ||
| GenBank mRNA | AK312075.1 | - | 3 | 234 | 82 | |
| BC140927.1 | - | 2 | 165 | 9056739 | 82 | |
| CA946421.1 | - | 2 | 118 | 82 | ||
| CD722957.1 | - | 3 | 219 | 82 | ||
Set Membership: |
|