Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs07292770_s1
See other ZNF273 GE Assays ›
Gene Symbol:
ZNF273
Gene Name:
zinc finger protein 273
Gene Aliases:
HZF9
Chromosome Location:
Chr.7: 64877603 - 64937316 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs07292770_s1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
4,430.00
Your Price
Online offer:
4430.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ZNF273
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001385643.1
NP_001372572.1
6
3372
109
NM_001385644.1
NP_001372573.1
8
2888
109
NM_001385645.1
NP_001372574.1
8
3506
109
NM_001385646.1
NP_001372575.1
4
2382
109
NM_001385649.1
NP_001372578.1
6
2754
109
NM_001385652.1
NP_001372581.1
5
2478
109
NM_001388021.1
NP_001374950.1
8
3445
109
NM_021148.3
NP_066971.2
4
2389
109
XM_024446635.2
XP_024302403.1
7
8636
109
XM_047419799.1
XP_047275755.1
3
2293
109
XM_047419800.1
XP_047275756.1
8
9425
109
XM_047419801.1
XP_047275757.1
7
8360
109
XM_047419802.1
XP_047275758.1
7
8542
109
XM_047419803.1
XP_047275759.1
8
9387
109
XM_047419804.1
XP_047275760.1
8
9381
109
XM_047419805.1
XP_047275761.1
7
8546
109
XM_047419806.1
XP_047275762.1
7
8597
109
XM_047419807.1
XP_047275763.1
8
3704
109
XM_047419808.1
XP_047275764.1
8
8449
109
XM_047419809.1
XP_047275765.1
9
4455
109
XM_047419810.1
XP_047275766.1
9
5636
109
XM_047419811.1
XP_047275767.1
5
7944
109
XM_047419812.1
XP_047275768.1
9
4257
109
XM_047419813.1
XP_047275769.1
9
4295
109
XM_047419814.1
XP_047275770.1
8
4212
109
XM_047419815.1
XP_047275771.1
7
3461
109
XM_047419816.1
XP_047275772.1
9
3549
109
XM_047419817.1
XP_047275773.1
9
3556
109
XM_047419818.1
XP_047275774.1
9
4251
109
XM_047419819.1
XP_047275775.1
8
3550
109
XM_047419820.1
XP_047275776.1
8
3460
109
XM_047419821.1
XP_047275777.1
8
3467
109
XM_047419822.1
XP_047275778.1
5
8562
109
GenBank mRNA
AL832810.1
-
1
2112
109
BC063818.1
-
5
2504
109
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Detect Genomic DNA
Recommended Assay
Homo sapiens
Both primers and probe map within a single exon
Matching Ambion siRNAs:
Click here to find siRNAs for Gene ZNF273
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
DNA binding
protein binding
zinc ion binding
metal ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.46.0-2026.03.11-1.0