| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013154.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013154.hg.1 |
ENST00000371471.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013154.hg.1 |
ENST00000395971.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013154.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR20013154.hg.1 |
TC20000956.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000437222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000437222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000437222.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000166422.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166422.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166422.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR2000166422.hg.1 |
TC2000009504.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR2000166422.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166422.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR2000166422.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000302342.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000371471.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000431687.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000437222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000437222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000437222.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
ENST00000540425.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC2000009504.hg.1 |
NM_006526.2 |
Refseq |