Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Hs99999141_s1
          See other JUN GE Assays ›
          Gene Symbol:
          JUN
          Gene Name:
          Jun proto-oncogene, AP-1 transcription factor subunit
          Gene Aliases:
          AP-1, AP1, c-Jun, cJUN, p39
          Chromosome Location:
          Chr.1: 58780791 - 58784047 on Build GRCh38
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          Assay ID Hs99999141_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (2)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: JUN
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_002228.4 NP_002219.1 1 1763 64
          GenBank mRNA BC002646.2 - 1 1732 64
          BC006175.1 - 1 1222 64
          BC009874.2 - 1 488 64
          BC068522.1 - 1 1762 64
          BT019759.1 - 1 786 64
          CR541724.1 - 1 786 64
          CR541752.1 - 1 786 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as Clariom D transcript cluster(s) TC0100014349.hg.1
          This assay targets the same location as HTA transcript cluster(s) TC01002708.hg.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Human PDGF Pathway Human Erythropoietin Pathway Human IL-2 Pathway Human TNFR2 Pathway Human Signal Transduction Pathways Human EGF Pathway Human IL-1 Pathway Human Toll-Like Receptors Pathway Human TNFR1 Pathway Human BMP Pathway Human Transcription Factors (non-Hox) Human TNF Superfamily Pathway HUMAN MOLECULAR MECHANISMS OF CANCER HUMAN FAS SIGNALING HUMAN INSULIN RECEPTOR PATHWAY HUMAN PI3K SIGNALING IN B-LYMPHOCYTE HUMAN 14-3-3 INDUCED INTRACELLULAR SIGNALING HUMAN GPCR PATHWAY HUMAN JAK-STAT PATHWAY HUMAN RHO FAMILY GTPASES HUMAN IL-3 PATHWAY HUMAN ILK SIGNALING HUMAN MELATONIN SIGNALING HUMAN RAS PATHWAY Inventoried Human_TNFR2_Pathway Human_EGF_Pathway Human_PDGF_Pathway Human_Toll-Like_Receptors_Pathway Human_IL-1_Pathway Human_BMP_Pathway Human_TNFR1_Pathway Human_Erythropoietin_Pathway Human_Signal_Transduction_Pathways Human_IL-2_Pathway Human_TNF_Superfamily_Pathway Human_Transcription_Factors_(non-Hox) Human_IL-3_Pathway Human_PI3K_Signaling_in_B-Lymphocyte Human_14-3-3_Induced_Intracellular_Signaling Human_Melatonin_Signaling Human_ILK_Signaling Human_GPCR_Pathway Human_Fas_Signaling Human_Molecular_Mechanisms_of_Cancer Human_Insulin_Receptor_Pathway Human_Ras_Pathway Human_Rho_Family_GTPases Human_JAK-STAT_Pathway Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene JUN

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          angiogenesis
          microglial cell activation
          liver development
          positive regulation of endothelial cell proliferation
          outflow tract morphogenesis
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          apoptotic process
          transforming growth factor beta receptor signaling pathway
          JNK cascade
          cell proliferation
          negative regulation of cell proliferation
          response to xenobiotic stimulus
          positive regulation of epithelial cell migration
          release from viral latency
          monocyte differentiation
          axon regeneration
          response to endoplasmic reticulum stress
          leading edge cell differentiation
          response to muscle stretch
          regulation of cell proliferation
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of DNA binding
          negative regulation of neuron apoptotic process
          negative regulation by host of viral transcription
          positive regulation by host of viral transcription
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fibroblast proliferation
          response to steroid hormone
          regulation of cell cycle
          SMAD protein signal transduction
          eyelid development in camera-type eye
          positive regulation of ERK1 and ERK2 cascade
          cellular response to calcium ion
          cellular response to anisomycin
          integrated stress response signaling
          positive regulation of pri-miRNA transcription from RNA polymerase II promoter
          positive regulation of vascular smooth muscle cell proliferation
          positive regulation of DNA-templated transcription, initiation
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          RNA binding
          GTPase activator activity
          protein binding
          enzyme binding
          ubiquitin protein ligase binding
          cAMP response element binding
          identical protein binding
          ubiquitin-like protein ligase binding
          RNA polymerase II sequence-specific DNA binding transcription factor binding
          R-SMAD binding
          general transcription initiation factor binding
          sequence-specific double-stranded DNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042580.hg.1 ENST00000371222.2 Ensembl
          Affymetrix Human Transcriptome Array 2.0 PSR01042580.hg.1 NM_002228.3 Refseq
          Affymetrix Human Transcriptome Array 2.0 PSR01042580.hg.1 TC01002708.hg.1 HTA TC
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay TC0100014349.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100207497.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100207497.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100207497.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100207497.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100207497.hg.1 TC0100014349.hg.1 Clariom D TC
          Affymetrix Human Clariom D Assay PSR0100207498.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100207498.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100207498.hg.1 ENST00000371222.3 Ensembl
          Affymetrix Human Clariom D Assay PSR0100207498.hg.1 NM_002228.3 Refseq
          Affymetrix Human Clariom D Assay PSR0100207498.hg.1 TC0100014349.hg.1 Clariom D TC

          Back To Top
           

          Citations (2)

          Epstein-Barr virus-encoded EBNA1 modulates the AP-1 transcription factor pathway in nasopharyngeal carcinoma cells and enhances angiogenesis in vitro.
          Authors
          O'Neil JD, Owen TJ, Wood VH, Date KL, Valentine R, Chukwuma MB, Arrand JR, Dawson CW, Young LS
          Journal
          J Gen Virol - Page No 2833 (Sat Nov 01 00:00:00 UTC 2008)
          Volume (Year)
          89 (2008)
          Product Usage
          -
          Comparative analysis of transcriptional profiling of CD3+, CD4+ and CD8+ T cells identifies novel immune response players in T-cell activation.
          Authors
          Wang M, Windgassen D, Papoutsakis ET
          Journal
          Bmc Genomics - Page No 225 (Tue Jan 01 00:00:00 UTC 2008)
          Volume (Year)
          9 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline