Search
Search

URI1| Gene Symbol: URI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005588703.2 | XP_005588760.1 | 9 - 10 | 1345 | 78 | |
| XM_005588706.2 | XP_005588763.1 | 8 - 9 | 1052 | 78 | ||
| XM_015441495.1 | XP_015296981.1 | 8 - 9 | 3751 | 78 | ||
| XM_015441496.1 | XP_015296982.1 | 8 - 9 | 1051 | 78 | ||
| XM_015441498.1 | XP_015296984.1 | 9 - 10 | 2285 | 78 | ||
| GenBank mRNA | AB173445.1 | - | 5 - 6 | 776 | 78 | |
Set Membership: |
|