Search
Search

LOC102140945| Gene Symbol: LOC102140945 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283192.1 | NP_001270121.1 | 2 - 3 | 57 | 84 | |
| XM_015452245.1 | XP_015307731.1 | 3 - 4 | 208 | 84 | ||
| GenBank mRNA | AF485815.1 | - | 2 - 3 | 57 | 84 | |
| DQ423377.1 | - | 2 - 3 | 57 | 84 | ||
Set Membership: |
|