Search
Search

LOC102121655| Gene Symbol: LOC102121655 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005576943.2 | XP_005577000.1 | 8 - 9 | 888 | 71 | |
| XM_005576944.1 | XP_005577001.1 | 8 - 9 | 907 | 71 | ||
| XM_005576946.2 | XP_005577003.1 | 8 - 9 | 876 | 71 | ||
| XM_005576947.2 | XP_005577004.1 | 7 - 8 | 772 | 71 | ||
| XM_005576949.2 | XP_005577006.1 | 8 - 9 | 810 | 71 | ||
| XM_005576950.2 | XP_005577007.1 | 8 - 9 | 888 | 71 | ||
| XM_005576951.2 | XP_005577008.1 | 7 - 8 | 761 | 71 | ||
| XM_015434311.1 | XP_015289797.1 | 8 - 9 | 898 | 71 | ||
| XM_015434312.1 | XP_015289798.1 | 8 - 9 | 886 | 71 | ||
| XM_015434313.1 | XP_015289799.1 | 8 - 9 | 819 | 71 | ||
| XM_015434314.1 | XP_015289800.1 | 4 - 5 | 617 | 71 | ||
Set Membership: |
|