Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00431617_m1
          See other ACADS GE Assays ›
          Gene Symbol:
          Acads
          Gene Name:
          acyl-Coenzyme A dehydrogenase, short chain
          Gene Aliases:
          AI196007, Bcd-1, Bcd1, Hdlq8, SCAD
          Chromosome Location:
          Chr.5: 115110299 - 115119346 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00431617_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Acads
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007383.3 NP_031409.2 2 - 3 284 57
          GenBank mRNA AK155361.1 - 2 - 3 263 57
          AK169428.1 - 2 - 3 221 57
          BC016259.1 - 2 - 3 251 4913813 57
          BC055063.1 - 2 - 3 225 57
          BY077302.1 - 2 - 3 285 57
          L11163.1 - 2 - 3 208 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500003088.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Acads

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          metabolic process
          fatty acid beta-oxidation using acyl-CoA dehydrogenase
          butyrate catabolic process
          protein homotetramerization
          oxidation-reduction process
          fatty-acyl-CoA binding
          acyl-CoA dehydrogenase activity
          butyryl-CoA dehydrogenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-CH group of donors
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24007909 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24007907 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24007908 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24007906 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 L11163.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 AK169428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 BC016259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 BC055063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 NM_007383.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 L11163.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 AK169428.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 BC016259.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 BC055063.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P367745 NM_007383.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 486659 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 215768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 569543 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393126 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173630 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133475 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146299 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 L11163.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 AK169428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 BC016259.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 BC055063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 NM_007383.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 L11163.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 AK169428.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 BC016259.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 BC055063.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P367745 NM_007383.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 L11163.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 AK169428.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 BC016259.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 BC055063.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 NM_007383.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 L11163.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 AK169428.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 BC016259.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 BC055063.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P367745 NM_007383.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007927 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007923 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24007907 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146299 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01194159 - -
          Whole Mouse Genome G4122A A_52_P367745 L11163.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367745 AK169428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367745 BC016259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367745 BC055063.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367745 NM_007383.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367745 L11163.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367745 AK169428.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367745 BC016259.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367745 BC055063.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P367745 NM_007383.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138684 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146299 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04247952 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04621857 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04763628 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04763630 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24007907 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24007909 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24007908 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24007906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24007898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007888 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04119847 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007906 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04783479 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04375380 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04119858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04388310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007911 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04119848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04934781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04119845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04621860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04486005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04783478 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007909 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04119857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04783477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007918 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04697201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04821801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04119852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007883 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007927 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007903 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04621863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P24007917 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10532935 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532934 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532929 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532930 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532928 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532936 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532933 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532927 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532931 - -
          Affymetrix Mouse Gene 1.0 ST Array 10532932 - -
          Affymetrix HT MG-430 PM Array Plate 1460216_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10532926 - -
          Mouse Genome 430 2.0 Array 1460216_PM_at - -
          Mouse Genome 430 2.0 Array 1460216_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11409_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11409_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0500027455.mm.1 TC0500003088.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500027455.mm.1 NM_007383.3 Refseq
          Mouse Expression Set 430 1460216_PM_at - -
          Mouse Expression Set 430 1460216_at - -
          Murine Genome U74v2 Set 161469_r_at - -
          Murine Genome U74v2 Set 103401_at - -
          Murine Genome U74v2 Set 103401_PM_at - -
          Murine Genome U74v2 Set 161469_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1460216_PM_at - -
          Mouse Genome 430A 2.0 Array 1460216_at - -
          Affymetrix HT Mouse Genome 430A Array 1460216_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1460216_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 550787 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 110746 - -
          Illumina mouse-6 v1.0 expression beadchip 110746 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000110746 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1256725 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256725 - -
          mouse-6 v1.1 (Illumina) scl26170.7.4_26-S - -
          Illumina Sentrix mouse-6.v1 bead array 110746 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl26170.7.4_26-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 110746 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2806676 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1256725 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2806676 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256725 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline