Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00431795_g1
          See other AMH GE Assays ›
          Gene Symbol:
          Amh
          Gene Name:
          anti-Mullerian hormone
          Gene Aliases:
          MIS
          Chromosome Location:
          Chr.10: 80805021 - 80807679 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00431795_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Amh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007445.2 NP_031471.2 4 - 5 811 73
          GenBank mRNA AY911505.1 - 4 - 5 811 73
          BC150154.1 - 4 - 5 810 73
          BC150477.1 - 4 - 5 810 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000872.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Amh

          Gene Ontology Categories:

          Function(s) Process(es)

          preantral ovarian follicle growth
          urogenital system development
          Mullerian duct regression
          cell-cell signaling
          positive regulation of gene expression
          positive regulation of NF-kappaB transcription factor activity
          negative regulation of ovarian follicle development
          receptor binding
          transforming growth factor beta receptor binding
          hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285630 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285631 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285634 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285629 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285628 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285632 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062953 NM_007445.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2062953 AY911505.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 81691 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062953 NM_007445.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2062953 AY911505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230075 NM_007445.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230075 AY911505.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230075 BC150154.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P230075 BC150477.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285631 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285627 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285626 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27285624 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07123677 - -
          Whole Mouse Genome G4122A A_51_P230075 NM_007445.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230075 AY911505.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230075 BC150154.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P230075 BC150477.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105618 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08415416 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08415439 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285628 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285629 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285632 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285630 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285634 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27285631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27285558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08415416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285633 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285622 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08415448 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08415418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27285623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08415449 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364906 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364907 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364908 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364904 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364905 - -
          Affymetrix HT MG-430 PM Array Plate 1450573_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10364903 - -
          Mouse Genome 430 2.0 Array 1450573_PM_at - -
          Mouse Genome 430 2.0 Array 1450573_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11705_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11705_at - -
          Affymetrix Mouse Clariom S Array PSR1000006682.mm.1 NM_007445.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006682.mm.1 NM_007445.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000006682.mm.1 TC1000000872.mm.1 MTA TC
          Mouse Expression Set 430 1450573_at - -
          Mouse Expression Set 430 1450573_PM_at - -
          Murine Genome U74v2 Set 101765_at - -
          Murine Genome U74v2 Set 101765_PM_at - -
          Mouse Genome 430A 2.0 Array 1450573_PM_at - -
          Mouse Genome 430A 2.0 Array 1450573_at - -
          Affymetrix HT Mouse Genome 430A Array 1450573_at - -
          Affymetrix HT Mouse Genome 430A Array 1450573_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 603816 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4730286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004730286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2768691 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2768691 - -
          mouse-6 v1.1 (Illumina) scl38674.5.1_203-S - -
          Illumina Sentrix mouse-6.v1 bead array 4730286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38674.5.1_203-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4730286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2768691 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2768691 - -

          Back To Top
           

          Citations (1)

          Effects of gestational diethylstilbestrol treatment on male and female gonads during early embryonic development.
          Authors
          Ikeda Y, Tanaka H, Esaki M
          Journal
          Endocrinology - Page No 3970 (Fri Aug 01 00:00:00 EDT 2008)
          Volume (Year)
          149 (2008)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline