Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00431814_m1
          See other APOA4 GE Assays ›
          Gene Symbol:
          Apoa4
          Gene Name:
          apolipoprotein A-IV
          Gene Aliases:
          Apoa-4
          Chromosome Location:
          Chr.9: 46240844 - 46243458 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00431814_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Apoa4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007468.2 NP_031494.2 2 - 3 484 72
          GenBank mRNA AK004856.1 - 2 - 3 267 72
          AK161535.1 - 2 - 3 267 72
          AK168687.1 - 2 - 3 268 72
          BC010769.1 - 2 - 3 277 4240796 72
          BC050149.1 - 2 - 3 242 30013747 72
          M64248.1 - 2 - 3 266 72
          M64249.1 - 2 - 3 266 72
          M64250.1 - 2 - 3 266 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900000560.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Apoa4

          Panther Classification:

          Molecular Function -

          apolipoprotein

          Gene Ontology Categories:

          Function(s) Process(es)

          innate immune response in mucosa
          cholesterol biosynthetic process
          transport
          lipid transport
          response to lipid hydroperoxide
          leukocyte cell-cell adhesion
          cholesterol metabolic process
          positive regulation of cholesterol esterification
          positive regulation of triglyceride catabolic process
          removal of superoxide radicals
          triglyceride catabolic process
          regulation of intestinal cholesterol absorption
          neuron projection regeneration
          regulation of cholesterol transport
          cholesterol efflux
          phospholipid efflux
          very-low-density lipoprotein particle remodeling
          high-density lipoprotein particle assembly
          negative regulation of plasma lipoprotein particle oxidation
          lipoprotein metabolic process
          cholesterol homeostasis
          hydrogen peroxide catabolic process
          reverse cholesterol transport
          multicellular organism lipid catabolic process
          positive regulation of fatty acid biosynthetic process
          phosphatidylcholine metabolic process
          positive regulation of lipoprotein lipase activity
          lipid homeostasis
          protein-lipid complex assembly
          triglyceride homeostasis
          copper ion binding
          lipid binding
          cholesterol binding
          antioxidant activity
          cholesterol transporter activity
          phosphatidylcholine binding
          protein homodimerization activity
          phosphatidylcholine-sterol O-acyltransferase activator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248725 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 738272 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 69024 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146239 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327491 - -
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327491 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478742 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478740 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26478739 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146239 - -
          Whole Mouse Genome G4122A A_51_P327491 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112122 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146239 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07920795 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07920804 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07920796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26478761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08038563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07999091 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07920788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07820490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478750 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478776 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07508174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07820488 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07920795 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478736 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07937283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07494810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478777 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08152440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07782367 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08063788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07937278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478788 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26478748 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10585014 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593176 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585011 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585012 - -
          Affymetrix Mouse Gene 1.0 ST Array 10585013 - -
          Affymetrix Mouse Gene 1.0 ST Array 10593175 - -
          Affymetrix HT MG-430 PM Array Plate 1417761_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1436504_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10593174 - -
          Affymetrix Mouse Gene 1.1 ST Array 10585010 - -
          Mouse Genome 430 2.0 Array 1417761_PM_at - -
          Mouse Genome 430 2.0 Array 1417761_at - -
          Mouse Genome 430 2.0 Array 1436504_x_at - -
          Mouse Genome 430 2.0 Array 1436504_PM_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11808_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11808_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900003930.mm.1 NM_007468.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003930.mm.1 NM_007468.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900003930.mm.1 TC0900000560.mm.1 MTA TC
          Mouse Expression Set 430 1436504_PM_x_at - -
          Mouse Expression Set 430 1417761_PM_at - -
          Mouse Expression Set 430 1417761_at - -
          Mouse Expression Set 430 1436504_x_at - -
          Murine Genome U74v2 Set 100078_at - -
          Murine Genome U74v2 Set 100078_PM_at - -
          Mouse Genome 430A 2.0 Array 1436504_x_at - -
          Mouse Genome 430A 2.0 Array 1417761_at - -
          Mouse Genome 430A 2.0 Array 1436504_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1417761_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1436504_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1436504_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1417761_at - -
          Affymetrix HT Mouse Genome 430A Array 1417761_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 929462 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102450537 - -
          MouseWG-6 v2 Expression BeadChip 4120451 - -
          Illumina mouse-6 v1.0 expression beadchip 102450537 - -
          Illumina mouse-6 v1.0 expression beadchip 4120451 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004120451 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2754104 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2754104 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1250349 - -
          mouse-6 v1.1 (Illumina) scl0011808.2_22-S - -
          mouse-6 v1.1 (Illumina) scl011808.3_227-S - -
          Illumina Sentrix mouse-6.v1 bead array 102450537 - -
          Illumina Sentrix mouse-6.v1 bead array 4120451 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0011808.2_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102450537 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120451 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2834123 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1250349 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2754104 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2834123 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline