Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00432058_m1
          See other OPN1SW GE Assays ›
          Gene Symbol:
          Opn1sw
          Gene Name:
          opsin 1 (cone pigments), short-wave-sensitive (color blindness, tritan)
          Gene Aliases:
          AW551857, Bcp
          Chromosome Location:
          Chr.6: 29376671 - 29388729 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00432058_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Opn1sw
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007538.3 NP_031564.1 4 - 5 1017 64
          GenBank mRNA AF190670.1 - 4 - 5 979 64
          BC026021.1 - 4 - 5 984 4511808 64
          BC058267.1 - 4 - 5 965 6489727 64
          U49720.1 - 4 - 5 919 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600001980.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Opn1sw

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          visual perception
          phototransduction
          protein-chromophore linkage
          response to stimulus
          signal transducer activity
          G-protein coupled receptor activity
          photoreceptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965886 BC026021.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965886 NM_007538.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 942089 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241688 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P163768 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965886 BC026021.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965886 NM_007538.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326102 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326105 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326112 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326107 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24326089 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P163768 - -
          Whole Mouse Genome G4122A A_52_P34185 BC026021.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P34185 NM_007538.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121476 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P163768 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05643615 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05643617 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05643623 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05643616 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05643609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24326133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05292725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05786687 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05825168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643609 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05786685 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643616 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05275350 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05292716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05292726 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05738228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05292718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05833790 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05381327 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05323660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05323657 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05290634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05323656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05833787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24326102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05643612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05786688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05275349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05290631 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10543596 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543592 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543598 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543599 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543594 - -
          Affymetrix Mouse Gene 1.0 ST Array 10543595 - -
          Affymetrix HT MG-430 PM Array Plate 1449132_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1418552_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10543591 - -
          Mouse Genome 430 2.0 Array 1418552_at - -
          Mouse Genome 430 2.0 Array 1418552_PM_at - -
          Mouse Genome 430 2.0 Array 1449132_at - -
          Mouse Genome 430 2.0 Array 1449132_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12057_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12057_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0600015535.mm.1 TC0600001980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015535.mm.1 NM_007538.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015535.mm.1 NM_007538.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015536.mm.1 TC0600001980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600015536.mm.1 NM_007538.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600015536.mm.1 NM_007538.3 Refseq
          Mouse Expression Set 430 1449132_at - -
          Mouse Expression Set 430 1418552_PM_at - -
          Mouse Expression Set 430 1418552_at - -
          Mouse Expression Set 430 1449132_PM_at - -
          Murine Genome U74v2 Set 99395_PM_at - -
          Murine Genome U74v2 Set 99395_at - -
          Mouse Genome 430A 2.0 Array 1449132_at - -
          Mouse Genome 430A 2.0 Array 1449132_PM_at - -
          Mouse Genome 430A 2.0 Array 1418552_at - -
          Mouse Genome 430A 2.0 Array 1418552_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418552_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449132_at - -
          Affymetrix HT Mouse Genome 430A Array 1418552_at - -
          Affymetrix HT Mouse Genome 430A Array 1449132_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 360138 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6420377 - -
          Illumina mouse-6 v1.0 expression beadchip 6420377 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006420377 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2593541 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2593541 - -
          mouse-6 v1.1 (Illumina) scl29212.6_52-S - -
          Illumina Sentrix mouse-6.v1 bead array 6420377 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl29212.6_52-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6420377 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2593541 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2893131 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2893131 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2593541 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl29212.6_52-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline