Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00432385_m1
          See other CCNF GE Assays ›
          Gene Symbol:
          Ccnf
          Gene Name:
          cyclin F
          Gene Aliases:
          CycF, Fbxo1
          Chromosome Location:
          Chr.17: 24223230 - 24251409 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00432385_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccnf
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007634.4 NP_031660.3 13 - 14 1590 62
          GenBank mRNA AK030298.1 - 1529 62
          AK032961.1 - 13 - 14 1516 62
          AK082485.1 - 13 - 14 1558 62
          AK159594.1 - 13 - 14 1591 62
          AK169263.1 - 13 - 14 1541 62
          AK214285.1 - 2 - 3 141 62
          BC037662.1 - 13 - 14 1536 5353169 62
          U20612.1 - 13 - 14 1528 62
          U20636.1 - 13 - 14 1645 62
          Z47766.1 - 13 - 14 1530 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001622.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccnf

          Panther Classification:

          Molecular Function -

          kinase activator

          Gene Ontology Categories:

          Function(s)

          re-entry into mitotic cell cycle
          placenta development
          cell cycle
          mitotic nuclear division
          negative regulation of centrosome duplication
          protein ubiquitination
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          cell division
          regulation of cell cycle

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096344 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096350 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096347 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096349 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096348 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096345 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039324 BC037662.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039324 AK169263.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039324 AK085183.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039324 AK159594.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2039324 AK030298.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462812 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 801242 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 272460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 846504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 399365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375409 AK032840.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375409 AK083213.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375409 AK085790.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375409 AK086802.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375409 BC062182.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375409 NM_029654.4 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 191527 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582674 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427103 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841432 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 413663 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 381976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468025 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 598149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 773224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 51412 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150757 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 92357 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174590 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119230 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039324 BC037662.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039324 AK169263.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039324 AK085183.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039324 AK159594.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2039324 AK030298.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328333 Z47766.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328333 U20612.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328333 U20636.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328333 BC037662.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P328333 AK169263.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096352 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096346 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096358 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096362 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096355 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096360 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096334 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096353 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31096335 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119230 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03293083 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174590 - -
          Whole Mouse Genome G4122A A_51_P328333 Z47766.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328333 U20612.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328333 U20636.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328333 BC037662.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P328333 AK169263.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108270 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104176 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174590 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13172213 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13172218 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13172217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13172216 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13172212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096347 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096350 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096344 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096346 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096345 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096348 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31096349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31096352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13172215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13217186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13545128 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13229422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096331 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13321786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096358 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13118009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13545127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096386 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13097513 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31096352 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448518 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448516 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448523 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448525 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448522 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448520 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448517 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448510 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448524 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448526 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448513 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448514 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448515 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448511 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448508 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448509 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448521 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448519 - -
          Affymetrix HT MG-430 PM Array Plate 1443807_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1422513_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10448506 - -
          Mouse Genome 430 2.0 Array 1443807_x_at - -
          Mouse Genome 430 2.0 Array 1422513_at - -
          Mouse Genome 430 2.0 Array 1443807_PM_x_at - -
          Mouse Genome 430 2.0 Array 1422513_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12449_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12449_at - -
          Affymetrix Murine Genome 6500A Array Msa14220_at - -
          Affymetrix Mouse Clariom S Array PSR1700014317.mm.1 NM_007634.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014317.mm.1 NM_007634.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014317.mm.1 TC1700001622.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014318.mm.1 NM_007634.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014318.mm.1 NM_007634.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014318.mm.1 TC1700001622.mm.1 MTA TC
          Mouse Expression Set 430 1422513_at - -
          Mouse Expression Set 430 1422513_PM_at - -
          Mouse Expression Set 430 1443807_x_at - -
          Mouse Expression Set 430 1443807_PM_x_at - -
          Murine Genome U74v2 Set 99073_at - -
          Murine Genome U74v2 Set 99073_PM_at - -
          Murine Genome U74v2 Set 141171_at - -
          Murine Genome U74v2 Set 141171_PM_at - -
          Mouse Genome 430A 2.0 Array 1422513_PM_at - -
          Mouse Genome 430A 2.0 Array 1422513_at - -
          Affymetrix HT Mouse Genome 430A Array 1422513_at - -
          Affymetrix HT Mouse Genome 430A Array 1422513_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1443807_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1443807_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 767163 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 106350133 - -
          Illumina mouse-6 v1.0 expression beadchip 6370288 - -
          Illumina mouse-6 v1.0 expression beadchip 106350133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006370288 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2722438 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219838 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2722438 - -
          mouse-6 v1.1 (Illumina) scl50213.19_604-S - -
          Illumina Sentrix mouse-6.v1 bead array 6370288 - -
          Illumina Sentrix mouse-6.v1 bead array 106350133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50213.19_604-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106350133 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6370288 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2722438 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1219838 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2722438 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline