Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00432423_m1
          See other CD79A GE Assays ›
          Gene Symbol:
          Cd79a
          Gene Name:
          CD79A antigen (immunoglobulin-associated alpha)
          Gene Aliases:
          Ig-alpha, Iga, Igalpha, Ly-54, Ly54, mb-1
          Chromosome Location:
          Chr.7: 24897511 - 24902197 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00432423_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cd79a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007655.3 NP_031681.2 1 - 2 76 89
          GenBank mRNA AK143605.1 - 1 - 2 77 89
          X13450.1 - 1 - 2 74 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000402.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cd79a

          Panther Classification:

          Molecular Function -

          immunoglobulin receptor superfamily

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          cell surface receptor signaling pathway
          B cell differentiation
          B cell proliferation
          B cell activation
          B cell receptor signaling pathway
          transmembrane signaling receptor activity
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P120987 AK143605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P120987 BC027633.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P120987 X13450.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P120987 NM_007655.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 614883 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 513965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 77611 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P120987 AK143605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P120987 BC027633.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P120987 X13450.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P120987 NM_007655.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2137496 BC027633.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P651316 AK143605.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P651316 BC027633.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P651316 X13450.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P651316 NM_007655.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007128 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007137 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25007165 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06142077 - -
          Whole Mouse Genome G4122A A_52_P651316 AK143605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651316 BC027633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651316 X13450.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P651316 NM_007655.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138674 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120987 AK143605.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120987 BC027633.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120987 X13450.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120987 NM_007655.3 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06172282 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06521814 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06521816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25007241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06465441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06611908 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06465452 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06572112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06223103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06465442 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06465447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06465457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007144 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06172281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06611902 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06572111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06250259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25007127 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10551029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551026 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551027 - -
          Affymetrix Mouse Gene 1.0 ST Array 10551028 - -
          Affymetrix Mouse Gene 1.1 ST Array 10551025 - -
          Mouse Genome 430 2.0 Array 1418830_at - -
          Mouse Genome 430 2.0 Array 1418830_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700002996.mm.1 TC0700000402.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002996.mm.1 NM_007655.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002996.mm.1 NM_007655.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002995.mm.1 TC0700000402.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700002995.mm.1 NM_007655.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700002995.mm.1 NM_007655.3 Refseq
          Mouse Expression Set 430 1418830_at - -
          Mouse Expression Set 430 1418830_PM_at - -
          Murine Genome U74v2 Set 102778_PM_at - -
          Murine Genome U74v2 Set 102778_at - -
          Mouse Genome 430A 2.0 Array 1418830_PM_at - -
          Mouse Genome 430A 2.0 Array 1418830_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 461806 - -
          ABI Mouse Genome Survey Microarray 898744 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3450563 - -
          Illumina mouse-6 v1.0 expression beadchip 3450563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246266 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246266 - -
          mouse-6 v1.1 (Illumina) scl32943.6.1_71-S - -
          Illumina Sentrix mouse-6.v1 bead array 3450563 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32943.6.1_71-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246266 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246266 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline