Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00432631_m1
          See other CORT GE Assays ›
          Gene Symbol:
          Cort
          Gene Name:
          cortistatin
          Gene Aliases:
          CST, PCST
          Chromosome Location:
          Chr.4: 149125191 - 149126741 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00432631_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cort
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007745.3 NP_031771.1 1 - 2 209 68
          GenBank mRNA AF013253.1 - 1 - 2 130 68
          AK013496.1 - 1 - 2 211 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004031.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cort

          Panther Classification:

          Molecular Function -

          neuropeptide

          Gene Ontology Categories:

          Function(s) Process(es)

          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          neuropeptide signaling pathway
          regulation of circadian sleep/wake cycle, sleep
          G-protein coupled receptor binding
          hormone activity
          neuropeptide hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2167323 NM_007745.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2167323 AK013496.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229196 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149989 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2167323 NM_007745.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2167323 AK013496.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P360241 NM_007745.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P360241 AF013253.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P360241 AK013496.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398784 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398790 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23398795 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149989 - -
          Whole Mouse Genome G4122A A_51_P360241 NM_007745.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P360241 AF013253.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P360241 AK013496.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P135918 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149989 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04092289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04092301 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04092284 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04092291 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03203840 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04092282 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P04092286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23398841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04092289 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490521 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04092290 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04092295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04092286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03490519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03551783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398779 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23398786 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10518559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518558 - -
          Affymetrix HT MG-430 PM Array Plate 1449820_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518557 - -
          Mouse Genome 430 2.0 Array 1449820_PM_at - -
          Mouse Genome 430 2.0 Array 1449820_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12854_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 12854_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400033376.mm.1 TC0400004031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033376.mm.1 NM_007745.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033373.mm.1 TC0400004031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033373.mm.1 NM_007745.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033375.mm.1 TC0400004031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033375.mm.1 NM_007745.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033374.mm.1 TC0400004031.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033374.mm.1 NM_007745.4 Refseq
          Mouse Expression Set 430 1449820_at - -
          Mouse Expression Set 430 1449820_PM_at - -
          Murine Genome U74v2 Set 99831_PM_at - -
          Murine Genome U74v2 Set 99831_at - -
          Mouse Genome 430A 2.0 Array 1449820_PM_at - -
          Mouse Genome 430A 2.0 Array 1449820_at - -
          Affymetrix HT Mouse Genome 430A Array 1449820_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1449820_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 516346 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 360008 AK014058.1 genbank_mrna_mouse
          MouseWG-6 v2 Expression BeadChip 360008 AK220542.1 genbank_mrna_mouse
          MouseWG-6 v2 Expression BeadChip 360008 NM_001113414.1 refseq_mrna_mouse_NM
          MouseWG-6 v2 Expression BeadChip 360008 NM_001113415.1 refseq_mrna_mouse_NM
          MouseWG-6 v2 Expression BeadChip 360008 NM_010096.3 refseq_mrna_mouse_NM
          MouseWG-6 v2 Expression BeadChip 2340092 - -
          Illumina mouse-6 v1.0 expression beadchip 360008 AK014058.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 360008 AK220542.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 360008 NM_001113414.1 refseq_mrna_mouse_NM
          Illumina mouse-6 v1.0 expression beadchip 360008 NM_001113415.1 refseq_mrna_mouse_NM
          Illumina mouse-6 v1.0 expression beadchip 360008 NM_010096.3 refseq_mrna_mouse_NM
          Illumina mouse-6 v1.0 expression beadchip 2340092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002340092 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360008 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213951 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226607 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213951 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226607 - -
          mouse-6 v1.1 (Illumina) scl0002856.1_27-S - -
          mouse-6 v1.1 (Illumina) scl23500.2.1_152-S - -
          Illumina Sentrix mouse-6.v1 bead array 360008 AK014058.1 genbank_mrna_mouse
          Illumina Sentrix mouse-6.v1 bead array 360008 AK220542.1 genbank_mrna_mouse
          Illumina Sentrix mouse-6.v1 bead array 360008 NM_001113414.1 refseq_mrna_mouse_NM
          Illumina Sentrix mouse-6.v1 bead array 360008 NM_001113415.1 refseq_mrna_mouse_NM
          Illumina Sentrix mouse-6.v1 bead array 360008 NM_010096.3 refseq_mrna_mouse_NM
          Illumina Sentrix mouse-6.v1 bead array 2340092 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23500.2.1_152-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002856.1_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2340092 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360008 AK014058.1 genbank_mrna_mouse
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360008 AK220542.1 genbank_mrna_mouse
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360008 NM_001113414.1 refseq_mrna_mouse_NM
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360008 NM_001113415.1 refseq_mrna_mouse_NM
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360008 NM_010096.3 refseq_mrna_mouse_NM
          MouseRef-8 v2 Expression BeadChip ILMN_1226607 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2955725 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2955725 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226607 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl23500.2.1_152-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline