Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00432769_m1
          See other CSTB GE Assays ›
          Gene Symbol:
          Cstb
          Gene Name:
          cystatin B
          Gene Aliases:
          AA960480, Stfb
          Chromosome Location:
          Chr.10: 78425670 - 78427622 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00432769_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cstb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007793.3 NP_031819.1 1 - 2 191 134
          GenBank mRNA AK002403.1 - 1 - 2 155 134
          AK151205.1 - 1 - 2 121 134
          BC056170.1 - 1 - 2 144 5713930 134
          BY091749.1 - 1 - 2 192 134

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000784.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cstb

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          adult locomotory behavior
          negative regulation of peptidase activity
          negative regulation of endopeptidase activity
          negative regulation of proteolysis
          protease binding
          endopeptidase inhibitor activity
          cysteine-type endopeptidase inhibitor activity
          peptidase inhibitor activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27274155 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27274153 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 AK002403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 AK151205.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 BC056170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 BY091749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 NM_007793.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 AK002403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 AK151205.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 BC056170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 BY091749.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P114910 NM_007793.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 165208 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P174049 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P119112 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149703 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 AK002403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 AK151205.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 BC056170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 BY091749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 NM_007793.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 AK002403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 AK151205.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 BC056170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 BY091749.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P114910 NM_007793.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 AK002403.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 AK151205.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 BC056170.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 BY091749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 NM_007793.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 AK002403.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 AK151205.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 BC056170.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 BY091749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P114910 NM_007793.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P7400 AK002403.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P7400 AK151205.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P7400 BC056170.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P7400 BY091749.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P7400 NM_007793.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274152 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27274163 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P174049 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149703 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P119112 - -
          Whole Mouse Genome G4122A A_52_P7400 AK002403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P7400 AK151205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P7400 BC056170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P7400 BY091749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P7400 NM_007793.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 AK002403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 AK151205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 BC056170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 BY091749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 NM_007793.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 AK002403.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 AK151205.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 BC056170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 BY091749.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P114910 NM_007793.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129562 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P119112 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P174049 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149703 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08371363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08321356 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08371364 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08371356 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08371361 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27274155 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27274153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27274174 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274163 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08876108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08297321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08642793 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08580914 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274132 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08892161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08672972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08596215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08371365 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274184 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274153 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08672976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08484298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27274161 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10364377 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364378 - -
          Affymetrix Mouse Gene 1.0 ST Array 10364376 - -
          Affymetrix HT MG-430 PM Array Plate 1422507_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1422506_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10364375 - -
          Mouse Genome 430 2.0 Array 1422506_PM_a_at - -
          Mouse Genome 430 2.0 Array 1422506_a_at - -
          Mouse Genome 430 2.0 Array 1422507_PM_at - -
          Mouse Genome 430 2.0 Array 1422507_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13014_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 13014_at - -
          Affymetrix Mouse Clariom S Array PSR1000005719.mm.1 NM_007793.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005719.mm.1 NM_007793.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005719.mm.1 TC1000000784.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005721.mm.1 NM_007793.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005721.mm.1 NM_007793.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005721.mm.1 TC1000000784.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005720.mm.1 NM_007793.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005720.mm.1 NM_007793.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005720.mm.1 TC1000000784.mm.1 MTA TC
          Mouse Expression Set 430 1422506_PM_a_at - -
          Mouse Expression Set 430 1422507_PM_at - -
          Mouse Expression Set 430 1422507_at - -
          Mouse Expression Set 430 1422506_a_at - -
          Murine Genome U74v2 Set 100581_at - -
          Murine Genome U74v2 Set 100581_PM_at - -
          Mouse Genome 430A 2.0 Array 1422506_a_at - -
          Mouse Genome 430A 2.0 Array 1422506_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1422507_PM_at - -
          Mouse Genome 430A 2.0 Array 1422507_at - -
          Affymetrix HT Mouse Genome 430A Array 1422507_at - -
          Affymetrix HT Mouse Genome 430A Array 1422507_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422506_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1422506_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 481863 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4560114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004560114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2700715 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2700715 - -
          mouse-6 v1.1 (Illumina) scl38726.3.1_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 4560114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl38726.3.1_35-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4560114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2700715 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2700715 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl38726.3.1_35-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline