Search
Search
| Gene Symbol: Edn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007902.2 | NP_031928.2 | 3 - 4 | 419 | 74 | |
| GenBank mRNA | BC037042.1 | - | 3 - 4 | 648 | 4512195 | 74 |
| X59556.1 | - | 3 - 4 | 410 | 74 | ||
Set Membership: |
|