Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433188_m1
          See other FABP2 GE Assays ›
          Gene Symbol:
          Fabp2
          Gene Name:
          fatty acid binding protein 2, intestinal
          Gene Aliases:
          Fabpi, I-FABP
          Chromosome Location:
          Chr.3: 122895072 - 122899506 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433188_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fabp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_007980.3 NP_032006.1 2 - 3 581 116
          GenBank mRNA AK007995.1 - 2 - 3 279 116
          AY523818.1 - 2 - 3 267 116
          AY523819.1 - 2 - 3 267 116
          BC013457.1 - 2 - 3 261 4218050 116
          BY707992.1 - 2 - 3 277 116
          M65034.1 - 2 - 3 275 116

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300001251.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fabp2

          Gene Ontology Categories:

          Function(s) Process(es)

          fatty acid metabolic process
          transport
          long-chain fatty acid transport
          intestinal absorption
          transporter activity
          long-chain fatty acid transporter activity
          fatty acid binding
          lipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313581 BC013457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313581 NM_007980.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313581 AK007995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313581 M65034.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313581 BC013457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313581 NM_007980.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313581 AK007995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P313581 M65034.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 35286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 780280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 42670 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P108151 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313581 BC013457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313581 NM_007980.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313581 AK007995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313581 M65034.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313581 BC013457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313581 NM_007980.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313581 AK007995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P313581 M65034.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313581 BC013457.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313581 NM_007980.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313581 AK007995.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313581 M65034.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313581 BC013457.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313581 NM_007980.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313581 AK007995.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P313581 M65034.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485111 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485120 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485100 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485110 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485124 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485097 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485106 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485109 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485093 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485103 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485089 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485104 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485101 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485117 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485114 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22485088 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P108151 - -
          Whole Mouse Genome G4122A A_51_P313581 BC013457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313581 NM_007980.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313581 AK007995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313581 M65034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313581 BC013457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313581 NM_007980.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313581 AK007995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P313581 M65034.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453013 AY523819.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453013 BC013457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453013 NM_007980.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453013 AK007995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P453013 AY523818.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108703 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P108151 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02861055 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02815818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02815820 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02826362 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02826367 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02826363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22485130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485102 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485114 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485104 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485124 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02763086 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02826377 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485136 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485079 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485127 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485135 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485120 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02826367 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485087 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485106 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485090 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485100 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485101 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485099 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485134 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02826368 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22485069 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02303550 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10495825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495823 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10495824 - -
          Affymetrix HT MG-430 PM Array Plate 1418438_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10495820 - -
          Mouse Genome 430 2.0 Array 1418438_PM_at - -
          Mouse Genome 430 2.0 Array 1418438_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14079_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14079_at - -
          Affymetrix Mouse Clariom S Array PSR0300010230.mm.1 NM_007980.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010230.mm.1 TC0300001251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010231.mm.1 NM_007980.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010231.mm.1 TC0300001251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010227.mm.1 NM_007980.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010227.mm.1 TC0300001251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010228.mm.1 NM_007980.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010228.mm.1 TC0300001251.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300010229.mm.1 NM_007980.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300010229.mm.1 TC0300001251.mm.1 MTA TC
          Mouse Expression Set 430 1418438_at - -
          Mouse Expression Set 430 1418438_PM_at - -
          Murine Genome U74v2 Set 97889_at - -
          Murine Genome U74v2 Set 97889_PM_at - -
          Mouse Genome 430A 2.0 Array 1418438_PM_at - -
          Mouse Genome 430A 2.0 Array 1418438_at - -
          Affymetrix HT Mouse Genome 430A Array 1418438_at - -
          Affymetrix HT Mouse Genome 430A Array 1418438_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 859800 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5050673 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050673 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2706493 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2706493 - -
          mouse-6 v1.1 (Illumina) scl22660.4.1_106-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050673 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl22660.4.1_106-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050673 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2706493 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2706493 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline