Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433357_m1
          See other FOLR2 GE Assays ›
          Gene Symbol:
          Folr2
          Gene Name:
          folate receptor 2 (fetal)
          Gene Aliases:
          FBP2, FR-P3, FR-beta, Folbp-2, Folbp2
          Chromosome Location:
          Chr.7: 101839988 - 101857190 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433357_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Folr2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008035.2 NP_032061.1 2 - 3 285 74
          GenBank mRNA BC022108.1 - 2 - 3 271 5102303 74
          CJ049116.1 - 2 - 3 286 74
          M64817.1 - 3 - 4 419 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003845.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Folr2

          Gene Ontology Categories:

          Function(s) Process(es)

          posttranslational protein targeting to membrane
          transport
          folic acid transport
          folic acid binding
          folic acid transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162671 M64817.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162671 NM_008035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162671 BC022108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162671 M64817.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162671 NM_008035.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P162671 BC022108.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 442955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166040 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 436700 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879344 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 147975 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 695582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 647929 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 406351 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 387073 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 778231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 60861 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 136012 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112268 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162671 M64817.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162671 NM_008035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162671 BC022108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162671 M64817.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162671 NM_008035.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P162671 BC022108.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162671 M64817.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162671 NM_008035.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162671 BC022108.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162671 M64817.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162671 NM_008035.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P162671 BC022108.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375794 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375749 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375753 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375781 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375747 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375775 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375807 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375751 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375768 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375769 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375776 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375762 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25375770 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112268 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06303478 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06303467 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06303454 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06303463 - -
          Whole Mouse Genome G4122A A_51_P162671 M64817.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162671 NM_008035.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162671 BC022108.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162671 M64817.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162671 NM_008035.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P162671 BC022108.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125115 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112268 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06009286 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06460664 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06009287 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06009289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06460668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25375688 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05917937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375772 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05926656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375786 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375778 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375766 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375801 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375767 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06137791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375798 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06023803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375791 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375799 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06106558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05917936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375784 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06214675 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05968155 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375753 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375783 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06250524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06358387 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375804 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375756 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06254988 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06137797 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06258368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375780 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06137792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375800 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25375776 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566033 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566029 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566027 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566031 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566028 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566030 - -
          Affymetrix Mouse Gene 1.0 ST Array 10566032 - -
          Affymetrix HT MG-430 PM Array Plate 1451648_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10566026 - -
          Mouse Genome 430 2.0 Array 1451648_a_at - -
          Mouse Genome 430 2.0 Array 1451648_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14276_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14276_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700032422.mm.1 NM_008035.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032422.mm.1 NM_008035.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032422.mm.1 TC0700003845.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032421.mm.1 NM_008035.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032421.mm.1 NM_008035.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032421.mm.1 TC0700003845.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700032420.mm.1 NM_008035.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032420.mm.1 NM_008035.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700032420.mm.1 TC0700003845.mm.1 MTA TC
          Mouse Expression Set 430 1451648_PM_a_at - -
          Mouse Expression Set 430 1451648_a_at - -
          Murine Genome U74v2 Set 104172_PM_at - -
          Murine Genome U74v2 Set 104172_at - -
          Mouse Genome 430A 2.0 Array 1451648_a_at - -
          Mouse Genome 430A 2.0 Array 1451648_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451648_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1451648_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 549296 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6020075 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006020075 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775030 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775030 - -
          mouse-6 v1.1 (Illumina) scl30960.6.1_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 6020075 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30960.6.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6020075 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2865285 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2775030 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2865285 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline