Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433361_m1
          See other FSHB GE Assays ›
          Gene Symbol:
          Fshb
          Gene Name:
          follicle stimulating hormone beta
          Gene Aliases:
          FSH, Fshbeta
          Chromosome Location:
          Chr.2: 107056140 - 107059651 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433361_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fshb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008045.2 NP_032071.1 2 - 3 223 75
          GenBank mRNA AK017593.1 - 2 - 3 224 75
          BC061159.1 - 2 - 3 223 30251083 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004284.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Mouse Estrogen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fshb

          Panther Classification:

          Molecular Function -

          peptide hormone

          Gene Ontology Categories:

          Function(s) Process(es)

          ovarian follicle development
          follicle-stimulating hormone signaling pathway
          regulation of osteoclast differentiation
          positive regulation of bone resorption
          Sertoli cell proliferation
          hormone activity
          follicle-stimulating hormone activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090884 AK017593.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090884 BC061159.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2090884 NM_008045.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 874314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177311 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 105284 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157310 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090884 AK017593.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090884 BC061159.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2090884 NM_008045.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P187052 AK017593.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P187052 BC061159.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P187052 NM_008045.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504099 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504091 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504096 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504073 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504076 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504098 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504080 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504085 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504059 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504083 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21504065 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157310 - -
          Whole Mouse Genome G4122A A_51_P187052 AK017593.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P187052 BC061159.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P187052 NM_008045.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125440 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157310 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01622720 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01622722 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01622710 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01622712 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01622726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21503984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21503990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21503995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21503985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21503992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21503987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504022 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21503998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21504012 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504076 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504026 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02221392 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504010 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01829036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01340298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01896400 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504074 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504105 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01888471 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504065 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504020 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504085 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504108 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504037 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504011 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01829041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01829045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504031 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01763978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01448892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504072 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504008 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504013 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504081 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504034 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504054 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504061 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504063 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01763972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01448897 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504016 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504098 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01204006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504001 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504023 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504070 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504018 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01522305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504025 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21504110 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01763977 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10485683 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485682 - -
          Affymetrix Mouse Gene 1.0 ST Array 10485684 - -
          Affymetrix HT MG-430 PM Array Plate 1450996_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10485681 - -
          Mouse Genome 430 2.0 Array 1450996_PM_at - -
          Mouse Genome 430 2.0 Array 1450996_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14308_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14308_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200036000.mm.1 NM_008045.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036000.mm.1 TC0200004284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200036001.mm.1 NM_008045.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200036001.mm.1 TC0200004284.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035999.mm.1 NM_008045.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035999.mm.1 TC0200004284.mm.1 MTA TC
          Mouse Expression Set 430 1450996_PM_at - -
          Mouse Expression Set 430 1450996_at - -
          Murine Genome U74v2 Set 101737_PM_at - -
          Murine Genome U74v2 Set 101737_at - -
          Mouse Genome 430A 2.0 Array 1450996_PM_at - -
          Mouse Genome 430A 2.0 Array 1450996_at - -
          Affymetrix HT Mouse Genome 430A Array 1450996_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450996_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 372125 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 7100592 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007100592 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2598788 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2598788 - -
          mouse-6 v1.1 (Illumina) scl18892.3_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 7100592 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl18892.3_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7100592 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2598788 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2598788 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline