Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433541_m1
          See other GCDH GE Assays ›
          Gene Symbol:
          Gcdh
          Gene Name:
          glutaryl-Coenzyme A dehydrogenase
          Gene Aliases:
          9030411L18, AI266902, D17825, GCD
          Chromosome Location:
          Chr.8: 84886387 - 84893921 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433541_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gcdh
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001044744.1 NP_001038209.2 8 - 9 1067 68
          NM_008097.2 NP_032123.3 7 - 8 1124 68
          GenBank mRNA AK078866.1 - 5 - 6 677 68
          AK079500.1 - 1 - 2 175 68
          AK165406.1 - 7 - 8 950 68
          BC061158.1 - 7 - 8 954 30314273 68
          U18992.1 - 7 - 8 905 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800002594.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gcdh

          Panther Classification:

          Molecular Function -

          dehydrogenase

          Gene Ontology Categories:

          Function(s) Process(es)

          acyl-CoA metabolic process
          metabolic process
          fatty acid oxidation
          fatty-acyl-CoA biosynthetic process
          oxidation-reduction process
          fatty-acyl-CoA binding
          acyl-CoA dehydrogenase activity
          glutaryl-CoA dehydrogenase activity
          oxidoreductase activity
          oxidoreductase activity, acting on the CH-CH group of donors
          flavin adenine dinucleotide binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986441 NM_008097.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986441 AK078866.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1986441 NM_001044744.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 692149 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 947481 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448534 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 117136 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131049 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986441 NM_008097.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986441 AK078866.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1986441 NM_001044744.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P260098 U18992.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022533 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022497 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022519 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022524 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022501 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022514 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022536 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022499 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022513 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022493 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022521 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022515 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022527 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022510 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022511 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022517 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022529 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022542 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022545 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022509 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022506 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022505 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022541 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022547 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022495 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022512 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022502 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022525 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022507 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022537 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01784312 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131049 - -
          Whole Mouse Genome G4122A A_51_P260098 U18992.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105353 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131049 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06742071 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06742067 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06742051 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07304641 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06742072 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06742073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07304640 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06742077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06742054 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07304635 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06742058 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07304638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022473 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022507 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022492 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022502 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022497 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022463 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022515 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022514 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022509 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022476 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022459 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022458 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06742072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022483 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022490 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022523 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022485 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06742078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022464 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06742066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022466 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06742052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022465 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022470 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06981414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022473 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022469 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07010251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022487 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022511 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06850128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06742055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022524 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06742075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06742054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022526 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022498 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022521 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022463 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022517 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10580246 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580244 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580239 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580241 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580234 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580240 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580235 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580237 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580243 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580245 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580238 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580242 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580236 - -
          Affymetrix HT MG-430 PM Array Plate 1448717_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10580233 - -
          Mouse Genome 430 2.0 Array 1448717_at - -
          Mouse Genome 430 2.0 Array 1448717_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 270076_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 270076_at - -
          Affymetrix Mouse Clariom S Array PSR0800019830.mm.1 NM_001044744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019830.mm.1 NM_001044744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019830.mm.1 NM_008097.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019830.mm.1 NM_008097.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019830.mm.1 TC0800002594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019829.mm.1 NM_001044744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019829.mm.1 NM_001044744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019829.mm.1 NM_008097.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019829.mm.1 NM_008097.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019829.mm.1 TC0800002594.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800019828.mm.1 NM_001044744.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019828.mm.1 NM_001044744.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019828.mm.1 NM_008097.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019828.mm.1 NM_008097.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800019828.mm.1 TC0800002594.mm.1 MTA TC
          Mouse Expression Set 430 1448717_at - -
          Mouse Expression Set 430 1448717_PM_at - -
          Murine Genome U74v2 Set 160194_PM_at - -
          Murine Genome U74v2 Set 162189_r_at - -
          Murine Genome U74v2 Set 162189_PM_r_at - -
          Murine Genome U74v2 Set 160194_at - -
          Mouse Genome 430A 2.0 Array 1448717_PM_at - -
          Mouse Genome 430A 2.0 Array 1448717_at - -
          Affymetrix HT Mouse Genome 430A Array 1448717_at - -
          Affymetrix HT Mouse Genome 430A Array 1448717_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 837334 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2190324 - -
          Illumina mouse-6 v1.0 expression beadchip 4280347 - -
          Illumina mouse-6 v1.0 expression beadchip 2190324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002190324 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004280347 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1228907 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222337 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222337 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1228907 - -
          mouse-6 v1.1 (Illumina) scl34554.11.1_42-S - -
          mouse-6 v1.1 (Illumina) scl000730.1_110-S - -
          Illumina Sentrix mouse-6.v1 bead array 2190324 - -
          Illumina Sentrix mouse-6.v1 bead array 4280347 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000730.1_110-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl34554.11.1_42-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2190324 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4280347 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1228907 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222337 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3136542 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1228907 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1222337 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3136542 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline