Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433610_s1
          See other GJA4 GE Assays ›
          Gene Symbol:
          Gja4
          Gene Name:
          gap junction protein, alpha 4
          Gene Aliases:
          AU020209, AW558810, Cnx37, Cx37, Gja-4
          Chromosome Location:
          Chr.4: 127311420 - 127314039 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433610_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gja4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008120.3 NP_032146.1 2 891 147
          GenBank mRNA AF216832.1 - 2 830 147
          AK143024.1 - 2 894 147
          BC056613.1 - 2 825 4971608 147

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gja4

          Panther Classification:

          Molecular Function -

          gap junction

          Gene Ontology Categories:

          Function(s)

          blood vessel development
          calcium ion transport
          cell communication
          cell-cell signaling

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23284658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23284657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23284659 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117352 AK143024.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117352 BC056613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117352 NM_008120.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117352 AF216832.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117352 AK143024.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117352 BC056613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117352 NM_008120.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P117352 AF216832.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5739 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 33945 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148675 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117352 AK143024.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117352 BC056613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117352 NM_008120.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117352 AF216832.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117352 AK143024.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117352 BC056613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117352 NM_008120.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P117352 AF216832.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P117352 AK143024.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P117352 BC056613.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P117352 NM_008120.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P117352 AF216832.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P117352 AK143024.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P117352 BC056613.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P117352 NM_008120.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P117352 AF216832.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284648 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284641 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284657 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284667 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284684 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284683 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284663 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284678 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284665 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284669 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284661 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148675 - -
          Whole Mouse Genome G4122A A_52_P117352 AK143024.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P117352 BC056613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P117352 NM_008120.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P117352 AF216832.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P117352 AK143024.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P117352 BC056613.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P117352 NM_008120.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P117352 AF216832.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110325 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148675 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03866085 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03866074 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03866080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03866083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03866086 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03866071 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03866070 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23284657 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23284658 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23284659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03705922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04063350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03974022 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284682 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03511820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03378309 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03566268 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284675 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03566267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03974015 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284627 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04099984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04043212 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10516483 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516482 - -
          Affymetrix HT MG-430 PM Array Plate 1416255_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10516481 - -
          Mouse Genome 430 2.0 Array 1416255_at - -
          Mouse Genome 430 2.0 Array 1416255_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14612_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14612_at - -
          Mouse Expression Set 430 1416255_PM_at - -
          Mouse Expression Set 430 1416255_at - -
          Murine Genome U74v2 Set 104729_PM_at - -
          Murine Genome U74v2 Set 104729_at - -
          Mouse Genome 430A 2.0 Array 1416255_PM_at - -
          Mouse Genome 430A 2.0 Array 1416255_at - -
          Affymetrix HT Mouse Genome 430A Array 1416255_at - -
          Affymetrix HT Mouse Genome 430A Array 1416255_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 832123 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6380452 - -
          Illumina mouse-6 v1.0 expression beadchip 6380452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737704 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737704 - -
          mouse-6 v1.1 (Illumina) scl23804.2_553-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23804.2_553-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2737704 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2737704 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl23804.2_553-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline