Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433651_s1
          See other GJB4 GE Assays ›
          Gene Symbol:
          Gjb4
          Gene Name:
          gap junction protein, beta 4
          Gene Aliases:
          Cnx30.3, Cx30.3, Gjb-4
          Chromosome Location:
          Chr.4: 127331418 - 127354141 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433651_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gjb4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008127.4 NP_032153.1 2 722 67
          GenBank mRNA AK132468.1 - 2 719 67
          BC064060.1 - 1 660 1498230 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003567.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gjb4

          Panther Classification:

          Molecular Function -

          gap junction

          Gene Ontology Categories:

          Function(s)

          cell communication
          sensory perception of smell
          olfactory behavior

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972381 BC064060.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1972381 NM_008127.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853443 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 104050 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905650 AK002285.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905650 AK004049.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905650 AK007337.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905650 AK019922.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905650 AK137906.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905650 AK147477.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905650 AK216319.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905650 BC024336.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905650 NM_023260.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 907904 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 100604 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 144865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740305 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 759760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176608 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115506 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972381 BC064060.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1972381 NM_008127.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P187121 BC064060.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P187121 AK076408.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P187121 NM_008127.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23284872 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115506 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00983080 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00983084 - -
          Whole Mouse Genome G4122A A_51_P187121 BC064060.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P187121 AK076408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P187121 NM_008127.3 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114325 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115506 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03452083 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03452073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03452080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03452084 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03452078 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03452082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23284850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284836 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03748828 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03748834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03748839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284878 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284862 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284861 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284842 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284866 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284830 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03300553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03416672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03300541 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23284879 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10516488 - -
          Affymetrix Mouse Gene 1.0 ST Array 10516489 - -
          Affymetrix HT MG-430 PM Array Plate 1422179_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10516487 - -
          Mouse Genome 430 2.0 Array 1422179_PM_at - -
          Mouse Genome 430 2.0 Array 1422179_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14621_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14621_at - -
          Affymetrix Mouse Clariom S Array PSR0400029038.mm.1 TC0400003567.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029038.mm.1 NM_008127.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0400029041.mm.1 TC0400003567.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400029041.mm.1 NM_008127.4 Refseq
          Mouse Expression Set 430 1422179_at - -
          Mouse Expression Set 430 1422179_PM_at - -
          Murine Genome U74v2 Set 99373_at - -
          Murine Genome U74v2 Set 99373_PM_at - -
          Mouse Genome 430A 2.0 Array 1422179_PM_at - -
          Mouse Genome 430A 2.0 Array 1422179_at - -
          Affymetrix HT Mouse Genome 430A Array 1422179_at - -
          Affymetrix HT Mouse Genome 430A Array 1422179_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 582409 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4850133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004850133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2600190 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2600190 - -
          mouse-6 v1.1 (Illumina) scl0014621.2_189-S - -
          Illumina Sentrix mouse-6.v1 bead array 4850133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0014621.2_189-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4850133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2600190 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2600190 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline