Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433832_m1
          See other NR3C1 GE Assays ›
          Gene Symbol:
          Nr3c1
          Gene Name:
          nuclear receptor subfamily 3, group C, member 1
          Gene Aliases:
          GR, Grl-1, Grl1
          Chromosome Location:
          Chr.18: 39410545 - 39519416 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433832_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nr3c1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008173.3 NP_032199.3 1 - 2 1249 68
          GenBank mRNA AK037733.1 - 1 - 2 1129 68
          AK155200.1 - 3 - 4 1331 68
          AK156323.1 - 3 - 4 1325 68
          BC008154.1 - 2 - 3 1557 68
          BC016462.1 - 3 - 4 1332 68
          BC129912.1 - 2 - 3 1227 40111802 68
          BC129913.1 - 2 - 3 1227 40111803 68
          DQ504162.1 - 1 - 2 1233 68
          HM236293.1 - 2 - 3 1209 68
          X04435.1 - 2 - 3 1227 68
          X13358.1 - 2 - 3 1209 68
          X13359.1 - 2 - 3 1209 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001243.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Nuclear Receptor Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nr3c1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of gluconeogenesis
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          glucocorticoid metabolic process
          regulation of glucose metabolic process
          positive regulation of glutamate secretion
          chromatin modification
          adrenal gland development
          negative regulation of synaptic plasticity
          regulation of glucocorticoid biosynthetic process
          regulation of cell proliferation
          maternal behavior
          glucocorticoid receptor signaling pathway
          negative regulation of vascular permeability
          steroid hormone mediated signaling pathway
          glucocorticoid mediated signaling pathway
          positive regulation of neuron apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          response to arsenic-containing substance
          chromatin-mediated maintenance of transcription
          mammary gland duct morphogenesis
          positive regulation of dendritic spine development
          positive regulation of cell growth involved in cardiac muscle cell development
          cellular response to steroid hormone stimulus
          cellular response to dexamethasone stimulus
          negative regulation of glucocorticoid mediated signaling pathway
          positive regulation of glucocorticoid receptor signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II regulatory region DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          transcription coactivator activity
          glucocorticoid receptor activity
          steroid binding
          protein binding
          transcription factor binding
          zinc ion binding
          lipid binding
          Hsp70 protein binding
          receptor tyrosine kinase binding
          heat shock protein binding
          protein complex binding
          glucocorticoid-activated RNA polymerase II transcription factor binding transcription factor activity
          hormone binding
          protein homodimerization activity
          sequence-specific DNA binding
          metal ion binding
          protein heterodimerization activity
          protein dimerization activity
          Hsp90 protein binding
          steroid hormone binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800009236.mm.1 NM_008173.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009236.mm.1 NM_008173.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009236.mm.1 TC1800001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009234.mm.1 NM_008173.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009234.mm.1 NM_008173.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009234.mm.1 TC1800001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009237.mm.1 NM_008173.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009237.mm.1 NM_008173.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009237.mm.1 TC1800001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009233.mm.1 NM_008173.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009233.mm.1 NM_008173.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009233.mm.1 TC1800001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009232.mm.1 NM_008173.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009232.mm.1 NM_008173.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009232.mm.1 TC1800001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009235.mm.1 NM_008173.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009235.mm.1 NM_008173.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009235.mm.1 TC1800001243.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800009238.mm.1 NM_008173.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009238.mm.1 NM_008173.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800009238.mm.1 TC1800001243.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 477364 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0