Search
Search

Cxcl1| Gene Symbol: Cxcl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008176.3 | NP_032202.1 | 3 - 4 | 333 | 74 | |
| GenBank mRNA | BC037997.1 | - | 3 - 4 | 300 | 74 | |
| BC132502.1 | - | 3 - 4 | 292 | 40130779 | 74 | |
| BC132504.1 | - | 3 - 4 | 292 | 40130781 | 74 | |
| BQ031102.1 | - | 1 - 2 | 74 | |||
| J04596.1 | - | 3 - 4 | 268 | 74 | ||
Set Membership: |
|