Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433930_m1
          See other SLC39A7 GE Assays ›
          Gene Symbol:
          Slc39a7
          Gene Name:
          solute carrier family 39 (zinc transporter), member 7
          Gene Aliases:
          AA408174, AI117660, AL024048, H-2Ke4, H2-Ke4, Ke-4, Ke4, Ring5, Zip7
          Chromosome Location:
          Chr.17: 34028266 - 34031690 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433930_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc39a7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077709.1 NP_001071177.1 7 - 8 1305 70
          NM_008202.2 NP_032228.2 6 - 7 1590 70
          GenBank mRNA AK087839.1 - 6 - 7 1285 70
          AK145956.1 - 5 - 6 1555 70
          AK159968.1 - 6 - 7 1466 70
          AK167344.1 - 5 - 6 1638 70
          AK212409.1 - 1 - 2 77 70
          AK214234.1 - 1 - 2 77 70
          BC115426.2 - 6 - 7 1444 40093613 70
          BC115427.1 - 6 - 7 1444 40093614 70
          KJ650218.1 - 6 - 7 1158 70
          M32010.1 - 6 - 7 1458 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700002811.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Slc39a7

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          zinc II ion transport
          metal ion transport
          transmembrane transport
          protein binding
          metal ion transmembrane transporter activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151613 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151614 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151612 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151615 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 BC115427.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 M32010.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 AK167344.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 BC115426.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 AK159968.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 BC115427.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 M32010.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 AK167344.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 BC115426.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P174016 AK159968.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324025 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 524100 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129604 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 BC115427.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 M32010.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 AK167344.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 BC115426.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 AK159968.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 BC115427.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 M32010.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 AK167344.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 BC115426.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P174016 AK159968.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 BC115427.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 M32010.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 AK167344.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 BC115426.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 AK159968.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 BC115427.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 M32010.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 AK167344.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 BC115426.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P174016 AK159968.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151601 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31151588 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129604 - -
          Whole Mouse Genome G4122A A_51_P174016 BC115427.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174016 M32010.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174016 AK167344.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174016 BC115426.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174016 AK159968.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174016 BC115427.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174016 M32010.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174016 AK167344.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174016 BC115426.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P174016 AK159968.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120223 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P113988 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129604 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13526468 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13118033 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13526474 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13184890 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13184891 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13526453 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13526481 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13526457 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151614 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151613 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151612 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31151615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31151549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13526477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13184891 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13526474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13526469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151609 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151613 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13118033 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31151605 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450124 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450125 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450123 - -
          Affymetrix HT MG-430 PM Array Plate 1416949_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450116 - -
          Mouse Genome 430 2.0 Array 1416949_PM_s_at - -
          Mouse Genome 430 2.0 Array 1416949_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14977_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14977_at - -
          Affymetrix Mouse Clariom S Array PSR1700017675.mm.1 NM_001077709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017675.mm.1 NM_001077709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017675.mm.1 NM_008202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017675.mm.1 NM_008202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017675.mm.1 TC1700002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017673.mm.1 NM_001077709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017673.mm.1 NM_001077709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017673.mm.1 NM_008202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017673.mm.1 NM_008202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017673.mm.1 TC1700002811.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017674.mm.1 NM_001077709.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017674.mm.1 NM_001077709.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017674.mm.1 NM_008202.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017674.mm.1 NM_008202.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017674.mm.1 TC1700002811.mm.1 MTA TC
          Mouse Expression Set 430 1416949_s_at - -
          Mouse Expression Set 430 1416949_PM_s_at - -
          Murine Genome U74v2 Set 103371_PM_at - -
          Murine Genome U74v2 Set 103371_at - -
          Mouse Genome 430A 2.0 Array 1416949_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1416949_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1416949_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1416949_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5570131 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005570131 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223755 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223755 - -
          mouse-6 v1.1 (Illumina) scl50031.6.1_11-S - -
          Illumina Sentrix mouse-6.v1 bead array 5570131 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50031.6.1_11-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5570131 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2603540 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3106053 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3032560 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2603540 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3106053 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3032560 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0