Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00433973_m1
          See other HOXD13 GE Assays ›
          Gene Symbol:
          Hoxd13
          Gene Name:
          homeobox D13
          Gene Aliases:
          Hox-4.8, spdh
          Chromosome Location:
          Chr.2: 74668310 - 74671599 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00433973_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hoxd13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008275.3 NP_032301.2 1 - 2 773 110
          GenBank mRNA AB221620.1 - 1 - 2 773 110
          AK077862.1 - 1 - 2 479 110
          BC109143.2 - 1 - 2 110
          BC109144.2 - 1 - 2 110
          X99291.1 - 1 - 2 773 110

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001091.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Hoxd13

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          multicellular organism development
          pattern specification process
          anterior/posterior pattern specification
          gland morphogenesis
          embryonic limb morphogenesis
          male genitalia development
          limb morphogenesis
          regulation of cell proliferation
          embryonic digit morphogenesis
          positive regulation of transcription from RNA polymerase II promoter
          embryonic hindgut morphogenesis
          prostate epithelial cord arborization involved in prostate glandular acinus morphogenesis
          morphogenesis of an epithelial fold
          branch elongation of an epithelium
          regulation of branching involved in prostate gland morphogenesis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338647 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338643 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591310 NM_008275.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591310 AK077862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591310 NM_008275.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P591310 AK077862.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 139361 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 177749 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52295 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146980 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591310 NM_008275.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591310 AK077862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591310 NM_008275.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P591310 AK077862.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P284571 X99291.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P284571 BC109144.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P284571 NM_008275.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P284571 AB221620.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P284571 AK077862.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338647 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338653 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338655 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338645 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338650 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21338649 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146980 - -
          Whole Mouse Genome G4122A A_52_P591310 NM_008275.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591310 AK077862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591310 NM_008275.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P591310 AK077862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284571 X99291.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284571 BC109144.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284571 NM_008275.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284571 AB221620.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P284571 AK077862.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P111229 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146980 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01889014 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338643 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338644 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338582 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338615 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01970152 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02187421 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338646 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02087758 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01889014 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338667 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338626 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338656 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10472948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472947 - -
          Affymetrix HT MG-430 PM Array Plate 1422239_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1440626_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10472946 - -
          Mouse Genome 430 2.0 Array 1440626_PM_at - -
          Mouse Genome 430 2.0 Array 1440626_at - -
          Mouse Genome 430 2.0 Array 1422239_at - -
          Mouse Genome 430 2.0 Array 1422239_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15433_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15433_at - -
          Affymetrix Mouse Clariom S Array PSR0200008810.mm.1 NM_008275.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008810.mm.1 TC0200001091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200008811.mm.1 NM_008275.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008811.mm.1 TC0200001091.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200008812.mm.1 NM_008275.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008812.mm.1 TC0200001091.mm.1 MTA TC
          Mouse Expression Set 430 1422239_at - -
          Mouse Expression Set 430 1422239_PM_at - -
          Mouse Expression Set 430 1440626_PM_at - -
          Mouse Expression Set 430 1440626_at - -
          Murine Genome U74v2 Set 102567_PM_at - -
          Murine Genome U74v2 Set 102567_at - -
          Mouse Genome 430A 2.0 Array 1422239_PM_at - -
          Mouse Genome 430A 2.0 Array 1422239_at - -
          Affymetrix HT Mouse Genome 430A Array 1422239_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422239_at - -
          Affymetrix HT Mouse Genome 430B Array 1440626_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1440626_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 826326 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2260603 - -
          Illumina mouse-6 v1.0 expression beadchip 2260603 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002260603 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2694339 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2694339 - -
          mouse-6 v1.1 (Illumina) scl015433.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 2260603 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl015433.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2260603 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2694339 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2694339 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline