Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434204_m1
          See other IL13 GE Assays ›
          Gene Symbol:
          Il13
          Gene Name:
          interleukin 13
          Gene Aliases:
          Il-13
          Chromosome Location:
          Chr.11: 53631323 - 53634702 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434204_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (8)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Il13
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008355.3 NP_032381.1 1 - 2 211 56
          GenBank mRNA M23504.1 - 1 - 2 212 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100002738.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Recommended Assay Mus musculus Mouse Chemokine Panel Mouse Immune Response Inventoried Mouse_Immune_Response Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Il13

          Gene Ontology Categories:

          Function(s) Process(es)

          microglial cell activation
          positive regulation of immunoglobulin production
          inflammatory response
          immune response
          regulation of proton transport
          positive regulation of B cell proliferation
          positive regulation of connective tissue growth factor production
          negative regulation of NAD(P)H oxidase activity
          response to nicotine
          positive regulation of tyrosine phosphorylation of Stat6 protein
          positive regulation of macrophage activation
          positive regulation of ion transport
          positive regulation of mast cell degranulation
          positive regulation of smooth muscle cell proliferation
          positive regulation of protein secretion
          positive regulation of release of sequestered calcium ion into cytosol
          cellular response to cytokine stimulus
          negative regulation of transforming growth factor beta production
          negative regulation of neuron death
          negative regulation of complement-dependent cytotoxicity
          positive regulation of pancreatic stellate cell proliferation
          negative regulation of endothelial cell apoptotic process
          cytokine activity
          cytokine receptor binding
          interleukin-13 receptor binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100024866.mm.1 NM_008355.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024866.mm.1 NM_008355.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100024866.mm.1 TC1100002738.mm.1 MTA TC

          Back To Top
           

          Citations (8)

          TLR9 signaling is essential for the innate NK cell response in murine cutaneous leishmaniasis.
          Authors
          Liese J, Schleicher U, Bogdan C
          Journal
          Eur J Immunol - Page No 3424 (Sat Dec 01 00:00:00 EST 2007)
          Volume (Year)
          37 (2007)
          Product Usage
          -
          Cytokines, signaling pathways, and effector molecules required for the control of Leishmania (Viannia) braziliensis in mice.
          Authors
          Rocha FJ, Schleicher U, Mattner J, Alber G, Bogdan C
          Journal
          Infect Immun - Page No 3823 (Wed Aug 01 00:00:00 EDT 2007)
          Volume (Year)
          75 (2007)
          Product Usage
          -
          IL-21 administration into the nostril alleviates murine allergic rhinitis.
          Authors
          Hiromura Y, Kishida T, Nakano H, Hama T, Imanishi J, Hisa Y, Mazda O
          Journal
          J Immunol - Page No 7157 (Thu Nov 15 00:00:00 EST 2007)
          Volume (Year)
          179 (2007)
          Product Usage
          -
          Administration of IL-33 induces airway hyperresponsiveness and goblet cell hyperplasia in the lungs in the absence of adaptive immune system.
          Authors
          Kondo Y, Yoshimoto T, Yasuda K, Futatsugi-Yumikura S, Morimoto M, Hayashi N, Hoshino T, Fujimoto J, Nakanishi K
          Journal
          Int Immunol - Page No 791 (Sun Jun 01 00:00:00 EDT 2008)
          Volume (Year)
          20 (2008)
          Product Usage
          -
          Inflammation triggers synaptic alteration and degeneration in experimental autoimmune encephalomyelitis.
          Authors
          Centonze D, Muzio L, Rossi S, Cavasinni F, De Chiara V, Bergami A, Musella A, D'Amelio M, Cavallucci V, Martorana A, Bergamaschi A, Cencioni MT, Diamantini A, Butti E, Comi G, Bernardi G, Cecconi F, Battistini L, Furlan R, Martino G
          Journal
          J Neurosci - Page No 3442 (Wed Mar 18 00:00:00 EDT 2009)
          Volume (Year)
          29 (2009)
          Product Usage
          -
          CD4(+)CD25 (+) Regulatory T Cells Suppress Th17-Responses in an Experimental Colitis Model.
          Authors
          Ogino H, Nakamura K, Ihara E, Akiho H, Takayanagi R
          Journal
          Dig Dis Sci - Page No (Thu Jun 03 00:00:00 EDT 2010)
          Volume (Year)
           (2010)
          Product Usage
          -
          C5 modulates airway hyperreactivity and pulmonary eosinophilia during enhanced respiratory syncytial virus disease by decreasing C3a receptor expression.
          Authors
          Melendi GA, Hoffman SJ, Karron RA, Irusta PM, Laham FR, Humbles A, Schofield B, Pan CH, Rabold R, Thumar B, Thumar A, Gerard NP, Mitzner W, Barnum SR, Gerard C, Kleeberger SR, Polack FP
          Journal
          J Virol - Page No 991 (Mon Jan 01 00:00:00 EST 2007)
          Volume (Year)
          81 (2007)
          Product Usage
          -
          IL-4(-/-) mice with lethal Mesocestoides corti infections--reduced Th2 cytokines and alternatively activated macrophages.
          Authors
          O'Connell AE, Kerepesi LA, Vandergrift GL, Herbert DR, VAN Winkle TJ, Hooper DC, Pearce EJ, Abraham D
          Journal
          Parasite Immunol - Page No 741 (Tue Dec 01 00:00:00 EST 2009)
          Volume (Year)
          31 (2009)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline