Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434548_m1
          See other ITIH3 GE Assays ›
          Gene Symbol:
          Itih3
          Gene Name:
          inter-alpha trypsin inhibitor, heavy chain 3
          Gene Aliases:
          AW108094, Intin3, Itih-3
          Chromosome Location:
          Chr.14: 30908572 - 30923755 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434548_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Itih3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008407.2 NP_032433.2 6 - 7 700 71
          GenBank mRNA AK146700.1 - 6 - 7 701 71
          BC015276.1 - 6 - 7 658 4212290 71
          X70393.1 - 6 - 7 657 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1400001661.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Itih3

          Panther Classification:

          Molecular Function -

          protease inhibitor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of peptidase activity
          hyaluronan metabolic process
          serine-type endopeptidase inhibitor activity
          peptidase inhibitor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996674 NM_008407.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996674 AK146700.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1996674 AW047821.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 790009 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 221046 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 546423 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 322540 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 230400 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375211 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 765793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 320482 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 142486 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P156720 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102173 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996674 NM_008407.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996674 AK146700.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1996674 AW047821.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P367100 BC015276.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P367100 X70393.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508722 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508731 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508736 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508743 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508726 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508720 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508741 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508729 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508748 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29508717 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08071242 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102173 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08071278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P156720 - -
          Whole Mouse Genome G4122A A_51_P367100 BC015276.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P367100 X70393.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114838 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P156720 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102173 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11186872 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11186869 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11186867 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11573278 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11186873 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11186868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11186871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508724 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508726 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29508710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11180844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508742 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11613783 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508744 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508692 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508726 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508721 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508711 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508741 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508683 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508719 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11746146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508685 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11613779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508716 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508746 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508700 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508679 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508718 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508702 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508697 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11764826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508695 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508735 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508738 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508681 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11431662 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11186868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508717 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11746147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29508729 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11180848 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10418446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418447 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418437 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418443 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418436 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418438 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10418435 - -
          Affymetrix HT MG-430 PM Array Plate 1449123_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10418434 - -
          Mouse Genome 430 2.0 Array 1449123_PM_at - -
          Mouse Genome 430 2.0 Array 1449123_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16426_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16426_at - -
          Affymetrix Mouse Clariom S Array PSR1400012575.mm.1 NM_008407.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012575.mm.1 NM_008407.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012575.mm.1 TC1400001661.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400012574.mm.1 NM_008407.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012574.mm.1 NM_008407.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1400012574.mm.1 TC1400001661.mm.1 MTA TC
          Mouse Expression Set 430 1449123_PM_at - -
          Mouse Expression Set 430 1449123_at - -
          Murine Genome U74v2 Set 100002_at - -
          Murine Genome U74v2 Set 100002_PM_at - -
          Mouse Genome 430A 2.0 Array 1449123_PM_at - -
          Mouse Genome 430A 2.0 Array 1449123_at - -
          Affymetrix HT Mouse Genome 430A Array 1449123_at - -
          Affymetrix HT Mouse Genome 430A Array 1449123_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 761206 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3830551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003830551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231336 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231336 - -
          mouse-6 v1.1 (Illumina) scl45772.22.1_79-S - -
          Illumina Sentrix mouse-6.v1 bead array 3830551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl45772.22.1_79-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3830551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2862623 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1231336 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2862623 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl45772.22.1_79-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline