| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0800016472.mm.1 |
NM_008455.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016472.mm.1 |
TC0800002144.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016473.mm.1 |
NM_008455.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016473.mm.1 |
TC0800002144.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016471.mm.1 |
NM_008455.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016471.mm.1 |
TC0800002144.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016476.mm.1 |
NM_008455.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016476.mm.1 |
TC0800002144.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0800016474.mm.1 |
NM_008455.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0800016474.mm.1 |
TC0800002144.mm.1 |
MTA TC |