Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434774_g1
          See other LTB GE Assays ›
          Gene Symbol:
          Ltb
          Gene Name:
          lymphotoxin B
          Gene Aliases:
          AI662801, LTbeta, Tnfc, Tnfsf3, p33
          Chromosome Location:
          Chr.17: 35194507 - 35196305 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434774_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ltb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008518.2 NP_032544.1 1 - 2 176 56
          GenBank mRNA BC064062.1 - 1 - 2 160 56
          BC114919.1 - 1 - 2 156 56
          U16985.1 - 1 - 2 161 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000673.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ltb

          Gene Ontology Categories:

          Function(s) Process(es)

          immune response
          gene expression
          skin development
          positive regulation of interleukin-12 biosynthetic process
          lymph node development
          cytokine activity
          tumor necrosis factor receptor binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158188 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158193 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158190 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158191 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158192 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158195 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158194 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P302358 U16985.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P302358 BC064062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P302358 BC114919.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P302358 NM_008518.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P302358 U16985.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P302358 BC064062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P302358 BC114919.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P302358 NM_008518.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 391803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 41793 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P302358 U16985.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P302358 BC064062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P302358 BC114919.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P302358 NM_008518.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P302358 U16985.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P302358 BC064062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P302358 BC114919.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P302358 NM_008518.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302358 U16985.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302358 BC064062.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302358 BC114919.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302358 NM_008518.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302358 U16985.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302358 BC064062.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302358 BC114919.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P302358 NM_008518.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31158183 - -
          Whole Mouse Genome G4122A A_51_P302358 U16985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302358 BC064062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302358 BC114919.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302358 NM_008518.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302358 U16985.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302358 BC064062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302358 BC114919.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P302358 NM_008518.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140577 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13231062 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13231047 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158188 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158191 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158190 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158194 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158195 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158193 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31158192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158195 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13199204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13231054 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158185 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13231061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13231065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158186 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13164356 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13516002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158200 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10444753 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10444754 - -
          Affymetrix HT MG-430 PM Array Plate 1419135_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10444752 - -
          Mouse Genome 430 2.0 Array 1419135_at - -
          Mouse Genome 430 2.0 Array 1419135_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16994_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16994_at - -
          Affymetrix Mouse Clariom S Array PSR1700006866.mm.1 NM_008518.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006866.mm.1 NM_008518.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006866.mm.1 TC1700000673.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006864.mm.1 NM_008518.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006864.mm.1 NM_008518.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006864.mm.1 TC1700000673.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006863.mm.1 NM_008518.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006863.mm.1 NM_008518.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006863.mm.1 TC1700000673.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700006865.mm.1 NM_008518.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006865.mm.1 NM_008518.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700006865.mm.1 TC1700000673.mm.1 MTA TC
          Mouse Expression Set 430 1419135_at - -
          Mouse Expression Set 430 1419135_PM_at - -
          Murine Genome U74v2 Set 102940_at - -
          Murine Genome U74v2 Set 102940_PM_at - -
          Mouse Genome 430A 2.0 Array 1419135_at - -
          Mouse Genome 430A 2.0 Array 1419135_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419135_at - -
          Affymetrix HT Mouse Genome 430A Array 1419135_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 450136 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3990672 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003990672 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1258283 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1258283 - -
          mouse-6 v1.1 (Illumina) scl50784.3.81_39-S - -
          Illumina Sentrix mouse-6.v1 bead array 3990672 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50784.3.81_39-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3990672 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1258283 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1258283 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline