Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00434880_m1
          See other SMCP GE Assays ›
          Gene Symbol:
          Smcp
          Gene Name:
          sperm mitochondria-associated cysteine-rich protein
          Gene Aliases:
          Mcs, Mcsp
          Chromosome Location:
          Chr.3: 92583866 - 92589024 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00434880_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smcp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008574.3 NP_032600.2 1 - 2 199 75
          GenBank mRNA AK077172.1 - 1 - 2 170 75
          BC055104.1 - 1 - 2 169 6773089 75
          BC061167.1 - 1 - 2 149 6771333 75
          BU936528.1 - 1 - 2 202 75
          M29603.1 - 1 - 2 172 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300002397.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the M29603.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smcp

          Gene Ontology Categories:

          Function(s)

          single fertilization
          binding of sperm to zona pellucida
          penetration of zona pellucida
          sperm motility

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 NM_008574.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 AK077172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 BC055104.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 BC061167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 BY456161.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 NM_008574.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 AK077172.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 BC055104.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 BC061167.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P392385 BY456161.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 666192 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 211248 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 730976 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 141125 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P130987 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 NM_008574.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 AK077172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 BC055104.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 BC061167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 BY456161.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 NM_008574.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 AK077172.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 BC055104.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 BC061167.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P392385 BY456161.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 NM_008574.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 AK077172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 BC055104.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 BC061167.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 BY456161.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 NM_008574.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 AK077172.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 BC055104.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 BC061167.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P392385 BY456161.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332153 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332166 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22332167 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P130987 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04989575 - -
          Whole Mouse Genome G4122A A_51_P392385 NM_008574.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P392385 AK077172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P392385 BC055104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P392385 BC061167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P392385 BY456161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P392385 NM_008574.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P392385 AK077172.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P392385 BC055104.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P392385 BC061167.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P392385 BY456161.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115872 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P130987 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02828594 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02828615 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02828599 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02828604 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02828603 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02828606 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02828596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332086 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332093 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332097 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22332083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332148 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332130 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332159 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332162 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332142 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02657871 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332129 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332171 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332177 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02828609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02828604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02583622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02657875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332161 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332176 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332143 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02828602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332168 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332154 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332133 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332144 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02828606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02657865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332123 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332126 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332125 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02828597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02828600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332138 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22332153 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10499908 - -
          Affymetrix Mouse Gene 1.0 ST Array 10499907 - -
          Affymetrix HT MG-430 PM Array Plate 1418358_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10499906 - -
          Mouse Genome 430 2.0 Array 1418358_PM_at - -
          Mouse Genome 430 2.0 Array 1418358_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17235_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17235_at - -
          Affymetrix Murine Genome 6500A Array Msa13420_at - -
          Affymetrix Mouse Clariom S Array PSR0300018590.mm.1 NM_008574.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018590.mm.1 TC0300002397.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018591.mm.1 NM_008574.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018591.mm.1 TC0300002397.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300018594.mm.1 NM_008574.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0300018594.mm.1 TC0300002397.mm.1 MTA TC
          Mouse Expression Set 430 1418358_PM_at - -
          Mouse Expression Set 430 1418358_at - -
          Murine Genome U74v2 Set 100891_PM_at - -
          Murine Genome U74v2 Set 100891_at - -
          Mouse Genome 430A 2.0 Array 1418358_at - -
          Mouse Genome 430A 2.0 Array 1418358_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418358_at - -
          Affymetrix HT Mouse Genome 430A Array 1418358_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 377563 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1190148 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190148 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2623212 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2623212 - -
          mouse-6 v1.1 (Illumina) scl21897.3.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190148 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21897.3.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190148 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2623212 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2623212 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline