Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00435456_g1
          See other ORM1 GE Assays ›
          Gene Symbol:
          Orm1
          Gene Name:
          orosomucoid 1
          Gene Aliases:
          Agp-1, Agp-2, Orm-1
          Chromosome Location:
          Chr.4: 63344566 - 63348162 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00435456_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Orm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008768.1 NP_032794.1 2 - 3 293 71
          GenBank mRNA BC012725.1 - 2 - 3 300 4162961 71
          CT010399.1 - 2 - 3 261 71
          M27008.1 - 2 - 3 293 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000718.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Orm1

          Gene Ontology Categories:

          Function(s)

          regulation of immune system process
          transport
          acute-phase response

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 M27008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 NM_008768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 BC012725.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 CT010399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 M12567.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 M27008.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 NM_008768.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 BC012725.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 CT010399.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P336325 M12567.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444946 AK007023.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444946 AK050707.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444946 AK078369.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444946 BC035338.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444946 XM_001478852.1 refseq_mrna_mouse_XM
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102469 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P150556 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P157486 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 M27008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 NM_008768.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 BC012725.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 CT010399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 M12567.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 M27008.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 NM_008768.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 BC012725.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 CT010399.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P336325 M12567.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 M27008.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 NM_008768.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 BC012725.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 CT010399.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 M12567.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 M27008.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 NM_008768.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 BC012725.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 CT010399.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P336325 M12567.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969783 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969796 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969798 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969808 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969812 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969804 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969805 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22969786 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102469 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P157486 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P150556 - -
          Whole Mouse Genome G4122A A_51_P336325 M27008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336325 NM_008768.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336325 BC012725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336325 CT010399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336325 M12567.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336325 M27008.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336325 NM_008768.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336325 BC012725.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336325 CT010399.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P336325 M12567.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122733 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P157486 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P150556 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102469 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03741968 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03741956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03741976 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03741946 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03741960 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03741951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969912 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22969765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03549700 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04079655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969831 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03741945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969780 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969820 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03937242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969784 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969815 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03741974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03532722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969786 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03537950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03741958 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03459731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969789 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22969782 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505441 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505440 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505442 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505444 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505443 - -
          Affymetrix HT MG-430 PM Array Plate 1451054_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10505438 - -
          Mouse Genome 430 2.0 Array 1451054_at - -
          Mouse Genome 430 2.0 Array 1451054_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18405_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18405_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400005160.mm.1 NM_008768.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005160.mm.1 TC0400000718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400005162.mm.1 NM_008768.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005162.mm.1 TC0400000718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400005159.mm.1 NM_008768.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005159.mm.1 TC0400000718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400005157.mm.1 NM_008768.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005157.mm.1 TC0400000718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400005158.mm.1 NM_008768.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005158.mm.1 TC0400000718.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400005161.mm.1 NM_008768.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400005161.mm.1 TC0400000718.mm.1 MTA TC
          Mouse Expression Set 430 1451054_at - -
          Mouse Expression Set 430 1451054_PM_at - -
          Murine Genome U74v2 Set 100437_g_at - -
          Murine Genome U74v2 Set 100436_at - -
          Murine Genome U74v2 Set 100437_PM_g_at - -
          Murine Genome U74v2 Set 100436_PM_at - -
          Mouse Genome 430A 2.0 Array 1451054_PM_at - -
          Mouse Genome 430A 2.0 Array 1451054_at - -
          Affymetrix HT Mouse Genome 430A Array 1451054_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451054_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1500563 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500563 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1226935 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1226935 - -
          mouse-6 v1.1 (Illumina) scl018405.4_2-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500563 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl018405.4_2-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500563 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1226935 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1226935 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl018405.4_2-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline