Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00435514_m1
          See other PCP2 GE Assays ›
          Gene Symbol:
          Pcp2
          Gene Name:
          Purkinje cell protein 2 (L7)
          Gene Aliases:
          L7, Pcp-2
          Chromosome Location:
          Chr.8: 3623369 - 3625545 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00435514_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001129803.1 NP_001123275.1 3 - 4 318 59
          NM_001129804.1 NP_001123276.1 3 - 4 290 59
          NM_008790.2 NP_032816.1 3 - 4 315 59
          GenBank mRNA AK003002.1 - 3 - 4 261 59
          AK160398.1 - 3 - 4 279 59
          BU531373.1 - 3 - 4 316 59
          CB522989.1 - 3 - 4 290 59
          EU164759.1 - 3 - 4 318 59
          M21532.1 - 3 - 4 285 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001613.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcp2

          Gene Ontology Categories:

          Function(s) Process(es)

          rhodopsin mediated signaling pathway
          positive regulation of GTPase activity
          guanyl-nucleotide exchange factor activity
          GTPase regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614880 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614879 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614882 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614881 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111399 NM_001195244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111399 AK011549.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111399 BU531373.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111399 CB522989.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2111399 EU164759.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 890616 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316427 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P114900 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111399 NM_001195244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111399 AK011549.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111399 BU531373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111399 CB522989.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2111399 EU164759.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614877 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25614851 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P114900 - -
          Whole Mouse Genome G4122A A_51_P448677 BU531373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448677 CB522989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448677 EU164759.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448677 M21532.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448677 AK003002.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643832 NM_001195244.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643832 AK011549.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643832 BU531373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643832 CB522989.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P643832 EU164759.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P124965 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P114900 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614883 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614882 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614880 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614881 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25614879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614910 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614768 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25614836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614855 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614857 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614841 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614861 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614844 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25614839 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10576754 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576756 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576755 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576750 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576752 - -
          Affymetrix HT MG-430 PM Array Plate 1424944_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419085_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1419084_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10576749 - -
          Mouse Genome 430 2.0 Array 1419084_PM_a_at - -
          Mouse Genome 430 2.0 Array 1419085_at - -
          Mouse Genome 430 2.0 Array 1419084_a_at - -
          Mouse Genome 430 2.0 Array 1419085_PM_at - -
          Mouse Genome 430 2.0 Array 1424944_at - -
          Mouse Genome 430 2.0 Array 1424944_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18545_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18545_at - -
          Affymetrix Murine Genome 6500A Array Msa1030_at - -
          Affymetrix Mouse Clariom S Array PSR0800013254.mm.1 NM_001129803.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013254.mm.1 NM_001129803.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013254.mm.1 NM_001129804.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013254.mm.1 NM_001129804.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013254.mm.1 NM_008790.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013254.mm.1 NM_008790.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800013254.mm.1 TC0800001613.mm.1 MTA TC
          Mouse Expression Set 430 1419084_a_at - -
          Mouse Expression Set 430 1424944_PM_at - -
          Mouse Expression Set 430 1419085_at - -
          Mouse Expression Set 430 1424944_at - -
          Mouse Expression Set 430 1419085_PM_at - -
          Mouse Expression Set 430 1419084_PM_a_at - -
          Murine Genome U74v2 Set 162452_PM_at - -
          Murine Genome U74v2 Set 102376_r_at - -
          Murine Genome U74v2 Set 102376_PM_r_at - -
          Murine Genome U74v2 Set 162452_at - -
          Mouse Genome 430A 2.0 Array 1419084_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1419084_a_at - -
          Mouse Genome 430A 2.0 Array 1419085_PM_at - -
          Mouse Genome 430A 2.0 Array 1419085_at - -
          Mouse Genome 430A 2.0 Array 1424944_PM_at - -
          Mouse Genome 430A 2.0 Array 1424944_at - -
          Affymetrix HT Mouse Genome 430A Array 1424944_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424944_at - -
          Affymetrix HT Mouse Genome 430A Array 1419085_at - -
          Affymetrix HT Mouse Genome 430A Array 1419084_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1419085_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1419084_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 550044 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1940537 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001940537 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2641457 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2641457 - -
          mouse-6 v1.1 (Illumina) scl35155.3_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 1940537 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35155.3_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940537 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2641456 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2641456 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline