Search
Search

Prkci| Gene Symbol: Prkci | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008857.3 | NP_032883.2 | 8 - 9 | 916 | 79 | |
| GenBank mRNA | AK144979.1 | - | 8 - 9 | 916 | 79 | |
| AK147710.1 | - | 8 - 9 | 888 | 79 | ||
| AK167411.1 | - | 8 - 9 | 885 | 79 | ||
| AK220517.1 | - | 8 - 9 | 940 | 79 | ||
| BC021630.1 | - | 8 - 9 | 787 | 3980628 | 79 | |
| D28577.1 | - | 8 - 9 | 711 | 79 | ||
Set Membership: |
|