Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00435905_m1
          See other PRG2 GE Assays ›
          Gene Symbol:
          Prg2
          Gene Name:
          proteoglycan 2, bone marrow
          Gene Aliases:
          MBP, mMBP, mMBP-1
          Chromosome Location:
          Chr.2: 84980461 - 84983632 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00435905_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008920.4 NP_032946.1 4 - 5 555 59
          GenBank mRNA AK010901.1 - 4 - 5 556 59
          AK156715.1 - 4 - 5 557 59
          BC109348.2 - 4 - 5 535 40055691 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200001227.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prg2

          Panther Classification:

          Molecular Function -

          extracellular matrix structural protein

          Gene Ontology Categories:

          Function(s) Process(es)

          defense response to nematode
          immune system process
          immune response
          negative regulation of interleukin-10 production
          positive regulation of interleukin-4 production
          regulation of cytokine biosynthetic process
          defense response to bacterium
          carbohydrate binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363400 NM_008920.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363400 AK010901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363400 AK156715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363400 BC109348.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363400 NM_008920.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363400 AK010901.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363400 AK156715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P363400 BC109348.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 873402 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 36920 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 131102 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124850 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363400 NM_008920.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363400 AK010901.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363400 AK156715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363400 BC109348.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363400 NM_008920.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363400 AK010901.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363400 AK156715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P363400 BC109348.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363400 NM_008920.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363400 AK010901.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363400 AK156715.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363400 BC109348.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363400 NM_008920.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363400 AK010901.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363400 AK156715.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P363400 BC109348.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395628 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395633 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395623 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395618 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395622 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395625 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395595 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395621 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21395634 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124850 - -
          Whole Mouse Genome G4122A A_51_P363400 NM_008920.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363400 AK010901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363400 AK156715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363400 BC109348.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363400 NM_008920.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363400 AK010901.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363400 AK156715.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P363400 BC109348.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101950 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110689 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124850 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01584363 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01584366 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01523483 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01584357 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01584358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21395688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01883200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01441267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01938376 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01576737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01888496 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395622 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01539707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01156183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395598 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01584359 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01814433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01814431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01539709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01523484 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395628 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01888497 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395588 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395610 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21395602 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10473405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473400 - -
          Affymetrix Mouse Gene 1.0 ST Array 10473403 - -
          Affymetrix HT MG-430 PM Array Plate 1422873_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10473399 - -
          Mouse Genome 430 2.0 Array 1422873_at - -
          Mouse Genome 430 2.0 Array 1422873_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19074_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19074_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200009825.mm.1 NM_008920.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009825.mm.1 NM_008920.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009825.mm.1 TC0200001227.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200009824.mm.1 NM_008920.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009824.mm.1 NM_008920.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200009824.mm.1 TC0200001227.mm.1 MTA TC
          Mouse Expression Set 430 1422873_PM_at - -
          Mouse Expression Set 430 1422873_at - -
          Murine Genome U74v2 Set 92728_PM_at - -
          Murine Genome U74v2 Set 161048_PM_at - -
          Murine Genome U74v2 Set 92728_at - -
          Murine Genome U74v2 Set 161048_at - -
          Mouse Genome 430A 2.0 Array 1422873_PM_at - -
          Mouse Genome 430A 2.0 Array 1422873_at - -
          Affymetrix HT Mouse Genome 430A Array 1422873_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422873_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 438119 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4070176 - -
          Illumina mouse-6 v1.0 expression beadchip 4070176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004070176 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2613878 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2613878 - -
          mouse-6 v1.1 (Illumina) scl20686.6.1_14-S - -
          Illumina Sentrix mouse-6.v1 bead array 4070176 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl20686.6.1_14-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4070176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2613878 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2613878 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl20686.6.1_14-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline