Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436299_s1
          See other RASL2-9 GE Assays ›
          Gene Symbol:
          Rasl2-9
          Gene Name:
          RAS-like, family 2, locus 9
          Gene Aliases:
          M2, Ran, Ran/M2, Rasl2-9-ps
          Chromosome Location:
          Chr.7: 5124942 - 5125950 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436299_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rasl2-9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009028.2 NP_033054.1 1 883 105
          GenBank mRNA AK006350.1 - 1 884 105
          BC049619.1 - 1 886 105
          BC061180.1 - 1 883 6771852 105
          BC099553.1 - 1 883 30917698 105
          L32752.1 - 1 899 105

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002186.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rasl2-9

          Panther Classification:

          Molecular Function -

          small GTPase

          Gene Ontology Categories:

          Function(s) Process(es)

          ribosomal subunit export from nucleus
          protein import into nucleus
          transport
          intracellular protein transport
          nucleocytoplasmic transport
          signal transduction
          small GTPase mediated signal transduction
          protein transport
          nucleotide binding
          GTPase activity
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24952261 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24952262 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119032 BC061180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119032 BC099553.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119032 BC138995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119032 BC145722.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119032 L32752.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119030 BC061180.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119030 BC099553.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119030 BC138995.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119030 BC145722.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119030 L32752.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 710678 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119032 BC061180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119032 BC099553.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119032 BC138995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119032 BC145722.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119032 L32752.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119030 BC061180.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119030 BC099553.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119030 BC138995.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119030 BC145722.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119030 L32752.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P470242 BC061180.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P470242 BC099553.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P470242 BC138995.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P470242 BC145722.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P470242 L32752.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24952283 - -
          Whole Mouse Genome G4122A A_51_P470242 BC061180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P470242 BC099553.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P470242 BC138995.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P470242 BC145722.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P470242 L32752.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136835 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05956936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06432034 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24952262 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24952261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24952223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05956936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06381447 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24952283 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559722 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559721 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559723 - -
          Affymetrix HT MG-430 PM Array Plate 1422656_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559720 - -
          Mouse Genome 430 2.0 Array 1422656_at - -
          Mouse Genome 430 2.0 Array 1422656_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19428_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 19428_at - -
          Affymetrix Mouse Clariom S Array PSR0700019987.mm.1 TC0700002186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019987.mm.1 NM_009028.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019987.mm.1 NM_009028.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019986.mm.1 TC0700002186.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019986.mm.1 NM_009028.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700019986.mm.1 NM_009028.2 Refseq
          Mouse Expression Set 430 1422656_PM_at - -
          Mouse Expression Set 430 1436490_x_at - -
          Mouse Expression Set 430 1422656_at - -
          Mouse Expression Set 430 1436490_PM_x_at - -
          Murine Genome U74v2 Set 161839_PM_f_at - -
          Murine Genome U74v2 Set 102821_PM_s_at - -
          Murine Genome U74v2 Set 161839_f_at - -
          Murine Genome U74v2 Set 102821_s_at - -
          Mouse Genome 430A 2.0 Array 1436490_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1422656_at - -
          Mouse Genome 430A 2.0 Array 1422656_PM_at - -
          Mouse Genome 430A 2.0 Array 1436490_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1422656_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422656_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 844157 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5050332 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005050332 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1229223 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1229223 - -
          mouse-6 v1.1 (Illumina) scl31785.1.4_192-S - -
          Illumina Sentrix mouse-6.v1 bead array 5050332 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31785.1.4_192-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5050332 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1229223 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1229223 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline