Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436439_m1
          See other CCL22 GE Assays ›
          Gene Symbol:
          Ccl22
          Gene Name:
          chemokine (C-C motif) ligand 22
          Gene Aliases:
          ABCD-1, DCBCK, MDC, Scya22
          Chromosome Location:
          Chr.8: 94745684 - 94751388 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436439_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccl22
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009137.2 NP_033163.1 2 - 3 228 78
          GenBank mRNA AF052505.1 - 2 - 3 320 78
          AF076596.1 - 2 - 3 209 78
          AF163476.1 - 2 - 3 198 78
          AJ238238.1 - 2 - 3 203 78
          AK155083.1 - 2 - 3 229 78
          AK155764.1 - 2 - 3 226 78
          AK156627.1 - 2 - 3 229 78
          AK170080.1 - 2 - 3 229 78
          BC012658.1 - 2 - 3 238 4192393 78

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001118.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Mouse Chemokine Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ccl22

          Panther Classification:

          Molecular Function -

          cytokine intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          chemotaxis
          inflammatory response
          immune response
          cytokine activity
          chemokine activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 AK156627.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 BC012658.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 AJ238238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 AK155083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 AK155764.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 AK156627.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 BC012658.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 AJ238238.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 AK155083.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P327996 AK155764.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 357840 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 292283 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 122876 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P177595 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 AK156627.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 BC012658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 AJ238238.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 AK155083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 AK155764.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 AK156627.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 BC012658.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 AJ238238.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 AK155083.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P327996 AK155764.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 AK156627.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 BC012658.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 AJ238238.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 AK155083.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 AK155764.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 AK156627.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 BC012658.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 AJ238238.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 AK155083.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P327996 AK155764.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079268 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26079274 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P177595 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06603486 - -
          Whole Mouse Genome G4122A A_51_P327996 AK156627.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327996 BC012658.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327996 AJ238238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327996 AK155083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327996 AK155764.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327996 AK156627.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327996 BC012658.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327996 AJ238238.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327996 AK155083.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P327996 AK155764.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P121013 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P177595 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06881215 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07408696 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07408699 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07408700 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07408694 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07408701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07408697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26079276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07408697 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07420646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07237099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079290 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07020449 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06884987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06701828 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06758806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07420649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07095738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06701812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07008568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07026713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07052082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06701817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07026712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079293 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26079284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07420653 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10574216 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574219 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574217 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574218 - -
          Affymetrix Mouse Gene 1.0 ST Array 10574215 - -
          Affymetrix HT MG-430 PM Array Plate 1417925_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10574213 - -
          Mouse Genome 430 2.0 Array 1417925_PM_at - -
          Mouse Genome 430 2.0 Array 1417925_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20299_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 20299_at - -
          Affymetrix Mouse Clariom S Array PSR0800008717.mm.1 NM_009137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008717.mm.1 NM_009137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008717.mm.1 TC0800001118.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800008716.mm.1 NM_009137.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008716.mm.1 NM_009137.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800008716.mm.1 TC0800001118.mm.1 MTA TC
          Mouse Expression Set 430 1417925_at - -
          Mouse Expression Set 430 1417925_PM_at - -
          Murine Genome U74v2 Set 102310_PM_at - -
          Murine Genome U74v2 Set 102310_at - -
          Mouse Genome 430A 2.0 Array 1417925_at - -
          Mouse Genome 430A 2.0 Array 1417925_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417925_at - -
          Affymetrix HT Mouse Genome 430A Array 1417925_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 828013 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6380086 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006380086 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220122 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220122 - -
          mouse-6 v1.1 (Illumina) scl33476.5_456-S - -
          Illumina Sentrix mouse-6.v1 bead array 6380086 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl33476.5_456-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6380086 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220122 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220122 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33476.5_456-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline