Search
Search

Mapk10| Gene Symbol: Mapk10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081567.1 | NP_001075036.1 | 4 - 5 | 797 | 88 | |
| NM_009158.2 | NP_033184.2 | 4 - 5 | 797 | 88 | ||
| GenBank mRNA | AB005665.1 | - | 1 - 2 | 180 | 88 | |
| AB096076.1 | - | 2 - 3 | 231 | 88 | ||
| AB096077.1 | - | 2 - 3 | 231 | 88 | ||
| AB096078.1 | - | 2 - 3 | 231 | 88 | ||
| AB096079.1 | - | 2 - 3 | 231 | 88 | ||
| AK036275.1 | - | 4 - 5 | 749 | 88 | ||
| AK042368.1 | - | 3 - 4 | 362 | 88 | ||
| AK082242.1 | - | 4 - 5 | 800 | 88 | ||
| AK163206.1 | - | 4 - 5 | 748 | 88 | ||
| BC026697.1 | - | 4 - 5 | 789 | 88 | ||
| BC046625.1 | - | 4 - 5 | 736 | 6494163 | 88 | |
| L35236.1 | - | 4 - 5 | 540 | 88 | ||
Set Membership: |
|