Search
Search
| Gene Symbol: Ptk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009184.2 | NP_033210.1 | 2 - 3 | 641 | 79 | |
| GenBank mRNA | BC125342.1 | - | 2 - 3 | 612 | 40129857 | 79 |
| BC125344.1 | - | 2 - 3 | 612 | 40129859 | 79 | |
| U16805.1 | - | 2 - 3 | 641 | 79 | ||
Set Membership: |
|