Search
Search

Slc17a1| Gene Symbol: Slc17a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170638.1 | NP_001164109.1 | 10 - 11 | 1293 | 113 | |
| NM_009198.3 | NP_033224.3 | 10 - 11 | 1385 | 113 | ||
| GenBank mRNA | AK164712.1 | - | 10 - 11 | 1295 | 113 | |
| BC013445.1 | - | 10 - 11 | 1326 | 4241557 | 113 | |
| X77241.1 | - | 10 - 11 | 1252 | 113 | ||
Set Membership: |
|