Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00436945_m1
          See other TFF1 GE Assays ›
          Gene Symbol:
          Tff1
          Gene Name:
          trefoil factor 1
          Gene Aliases:
          Bcei, PS2
          Chromosome Location:
          Chr.17: 31161395 - 31165065 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00436945_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tff1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009362.2 NP_033388.1 1 - 2 128 133
          GenBank mRNA AK008625.1 - 1 - 2 125 133
          AK008809.1 - 1 - 2 125 133
          AK008873.1 - 1 - 2 128 133
          AK010179.1 - 1 - 2 125 133
          AK131896.1 - 1 - 2 129 133
          AK160174.1 - 1 - 2 125 133
          BC117054.1 - 1 - 2 125 40126373 133
          BC117056.1 - 1 - 2 125 40126375 133
          Z21858.1 - 1 - 2 97 133

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001813.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Mouse Estrogen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tff1

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of cell proliferation
          response to wounding
          cell differentiation
          protein binding
          growth factor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112005 Z21858.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112005 NM_009362.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112005 AK008809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112005 AK131896.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021585 Z21858.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021585 AK160174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021585 NM_009362.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021585 AK008625.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2021585 AK008809.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144185 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021585 Z21858.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021585 AK160174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021585 NM_009362.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021585 AK008625.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2021585 AK008809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112005 Z21858.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112005 NM_009362.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112005 AK008809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112005 AK131896.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504884 BC117056.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504884 Z21858.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504884 AK160174.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504884 BC117054.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P504884 NM_009362.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138266 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138226 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138262 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31138227 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144185 - -
          Whole Mouse Genome G4122A A_51_P504884 BC117056.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504884 Z21858.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504884 AK160174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504884 BC117054.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P504884 NM_009362.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114023 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144185 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13295219 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13295224 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13295221 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13301194 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13295230 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13301182 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13295225 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13295231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31138239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13301180 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13486459 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13301194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138242 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138230 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13199288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13166091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138247 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138260 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13183362 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13330419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13301184 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13301182 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13301185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13295221 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138217 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138243 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138232 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13295224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13582327 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138270 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138271 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13166083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138264 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P13452027 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P31138224 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10449668 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449667 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449671 - -
          Affymetrix HT MG-430 PM Array Plate 1448156_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10449666 - -
          Mouse Genome 430 2.0 Array 1448156_PM_at - -
          Mouse Genome 430 2.0 Array 1448156_at - -
          Affymetrix Mouse Clariom S Array PSR1700016669.mm.1 NM_009362.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016669.mm.1 NM_009362.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016669.mm.1 TC1700001813.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700016668.mm.1 NM_009362.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016668.mm.1 NM_009362.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700016668.mm.1 TC1700001813.mm.1 MTA TC
          Mouse Expression Set 430 1448156_PM_at - -
          Mouse Expression Set 430 1448156_at - -
          Murine Genome U74v2 Set 160300_PM_at - -
          Murine Genome U74v2 Set 160300_at - -
          Murine Genome U74v2 Set 167147_PM_at - -
          Murine Genome U74v2 Set 167147_at - -
          Mouse Genome 430A 2.0 Array 1448156_at - -
          Mouse Genome 430A 2.0 Array 1448156_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 457999 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3850452 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003850452 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2830821 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2830821 - -
          mouse-6 v1.1 (Illumina) GI_6678306-S - -
          Illumina Sentrix mouse-6.v1 bead array 3850452 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip GI_6678306-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3850452 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2830821 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2830821 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline