Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00437164_m1
          See other TNNI3 GE Assays ›
          Gene Symbol:
          Tnni3
          Gene Name:
          troponin I, cardiac 3
          Gene Aliases:
          Tn1, cTnI
          Chromosome Location:
          Chr.7: 4518308 - 4522443 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00437164_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tnni3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009406.3 NP_033432.1 7 - 8 681 67
          GenBank mRNA AK169223.1 - 7 - 8 682 67
          BC061171.1 - 7 - 8 681 30300830 67
          BC100590.1 - 7 - 8 612 6787916 67
          U09181.1 - 5 - 6 553 67

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004636.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tnni3

          Panther Classification:

          Molecular Function -

          non-motor actin binding protein

          Gene Ontology Categories:

          Function(s) Process(es)

          vasculogenesis
          regulation of systemic arterial blood pressure by ischemic conditions
          cellular calcium ion homeostasis
          regulation of muscle contraction
          regulation of smooth muscle contraction
          striated muscle contraction
          heart development
          negative regulation of ATPase activity
          ventricular cardiac muscle tissue morphogenesis
          heart contraction
          cardiac muscle contraction
          actin binding
          calcium channel inhibitor activity
          protein kinase binding
          protein domain specific binding
          troponin C binding
          troponin T binding
          metal ion binding
          calcium-dependent protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 NM_009406.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 AK169223.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 BC061171.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 BC100590.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 U09181.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 NM_009406.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 AK169223.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 BC061171.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 BC100590.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P386983 U09181.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 330900 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 380339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 664112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 140223 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 37364 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P180566 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117445 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 NM_009406.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 AK169223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 BC061171.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 BC100590.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 U09181.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 NM_009406.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 AK169223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 BC061171.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 BC100590.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P386983 U09181.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P563123 NM_009406.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P563123 AK169223.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P563123 BC061171.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P563123 BC100590.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P563123 U09181.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 NM_009406.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 AK169223.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 BC061171.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 BC100590.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 U09181.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 NM_009406.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 AK169223.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 BC061171.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 BC100590.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P386983 U09181.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949264 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24949237 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P180566 - -
          Whole Mouse Genome G4122A A_51_P386983 NM_009406.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386983 AK169223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386983 BC061171.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386983 BC100590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386983 U09181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386983 NM_009406.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386983 AK169223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386983 BC061171.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386983 BC100590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P386983 U09181.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563123 NM_009406.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563123 AK169223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563123 BC061171.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563123 BC100590.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P563123 U09181.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131649 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P180566 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06455441 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05957084 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05957087 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05957086 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05957080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05957089 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24949241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06455441 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06216454 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06275920 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05957094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05957080 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24949264 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10559560 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559562 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559559 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559561 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10559567 - -
          Affymetrix HT MG-430 PM Array Plate 1422536_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10559558 - -
          Mouse Genome 430 2.0 Array 1422536_at - -
          Mouse Genome 430 2.0 Array 1422536_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21954_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21954_at - -
          Affymetrix Mouse Clariom S Array PSR0700019677.mm.1 TC0700004636.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700019677.mm.1 NM_009406.4 Refseq
          Mouse Expression Set 430 1422536_at - -
          Mouse Expression Set 430 1422536_PM_at - -
          Murine Genome U74v2 Set 100921_at - -
          Murine Genome U74v2 Set 100921_PM_at - -
          Mouse Genome 430A 2.0 Array 1422536_PM_at - -
          Mouse Genome 430A 2.0 Array 1422536_at - -
          Affymetrix HT Mouse Genome 430A Array 1422536_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422536_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 466641 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3163526 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3163526 - -
          mouse-6 v1.1 (Illumina) GI_40254606-A - -
          MouseRef-8 v2 Expression BeadChip ILMN_3163526 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3163526 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline