Search
Search
| Gene Symbol: Vdr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009504.4 | NP_033530.2 | 3 - 4 | 289 | 95 | |
| GenBank mRNA | AK030339.1 | - | 3 - 4 | 290 | 95 | |
| AK154647.1 | - | 3 - 4 | 264 | 95 | ||
| AK156789.1 | - | 3 - 4 | 305 | 95 | ||
| BC006716.1 | - | 3 - 4 | 255 | 3710866 | 95 | |
| CT010294.1 | - | 1 - 2 | 149 | 95 | ||
| D31969.1 | - | 3 - 4 | 257 | 95 | ||
Set Membership: |
|